Insulin-Transferrin-Selenium (ITS-G) Select Supplement (100X)
Gibco™

Insulin-Transferrin-Selenium (ITS-G) Select Supplement (100X)

Insulin-Transferrin-Selenium (ITS-G) Select Supplement is an animal-origin-free (AOF) basal medium supplement that is used to reduce the amount of FBS needed to culture cells.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A4000046502100 mL
A400004650110 mL
Catalog number A4000046502
Price (USD)
662.00
Each
Quantity:
100 mL
Price (USD)
662.00
Each
Insulin-Transferrin-Selenium (ITS-G) Select Supplement is an animal-origin-free (AOF) basal medium supplement that is used to reduce the amount of FBS needed to culture cells.

Gibco Insulin-Transferrin-Selenium (ITS-G) Select Supplement is an animal origin-free (AOF) basal medium supplement. The insulin and transferrin in the ITS-G Select formulation are produced in non-mammalian recombinant expression systems.

ITS-G is used as a basal medium supplement to reduce the amount of fetal bovine serum (FBS) needed to culture cells. Originally used with RPMI 1640 and Minimum Essential Medium (MEM), ITS-G is now routinely used with other basal media to support a wide variety of cell types in the presence of 2–4% FBS.

Insulin promotes glucose and amino acid uptake, lipogenesis, intracellular transport, and the synthesis of proteins and nucleic acids. Transferrin is an iron carrier and may also help reduce toxic levels of oxygen radicals and peroxide. Selenium, as sodium selenite, is a co-factor for glutathione peroxidase and other proteins and is used as an antioxidant in media.

The product formulation contains the following components:

  • Insulin (1000.0 mg/L)
  • Transferrin (550.0 mg/L)
  • Sodium selenite (0.67 mg/L)

cGMP manufacturing and quality system

ITS-G Select Supplement is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Concentration100 X
Culture TypeMammalian Cell Culture
Endotoxin Level≤ 1.00 EU/mL
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Osmolality260 to 320 mOsm/kg
Quantity100 mL
Shelf Life12 Months from Date of Manufacture
Shipping ConditionRoom Temperature
SourceAnimal Origin-free
FormLiquid
Product TypeCell Culture Supplement
SterilitySterile-filtered
Sterilization MethodSterile-filtered
pH7.8 to 9.0
Unit SizeEach

Frequently asked questions (FAQs)

What is the buffer composition of the various Insulin-Transferrin-Selenium (ITS) (100X) and Insulin-Transferrin-Selenium (ITS) Select (100X) supplements?

All ITS and ITS Select supplements are formulated in Earle's Balanced Salt Solution (EBSS).

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the difference between Insulin-Transferrin-Selenium (ITS) (100X) and Insulin-Transferrin-Selenium (ITS) Select (100X) supplements?

ITS Select supplements are animal origin-free (AOF) versions of the ITS supplements offered by Thermo Fisher Scientific.

ITS Select supplements use recombinant versions of both proteins in the formulation: insulin and transferrin.

All other versions of ITS, those without the Select designation, utilize recombinant insulin and a human-derived holo transferrin.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What are the individual concentrations of insulin, transferrin, and selenium in the Insulin-Transferrin-Selenium (ITS) (100X) supplements?

All ITS and ITS Select supplements have the following concentrations of their components:

Insulin: 1.0 g/L
Transferrin: 0.55 g/L
Selenium: 0.00067 g/L

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Can I store aliquots of Insulin-Transferrin-Selenium (ITS) (100X) supplements at -20 degrees C without compromising the integrity of the product?

We recommend storing all ITS and ITS Select supplements at 2-8 degrees C as there is a chance that the insulin will precipitate upon freezing and thawing.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Are all Insulin-Transferrin-Selenium (ITS) (100X) supplements offered by Thermo Fisher Scientific animal origin-free?

We offer both animal origin and animal origin-free versions of our ITS supplements. The animal origin-free product versions are referred to as ITS Select supplements.

Insulin-Transferrin-Selenium (ITS-G) (100X) (Cat. No. 41400045), Insulin-Transferrin-Selenium-Ethanolamine (ITS-X) (100X) (Cat. No. 51500056), and Insulin-Transferrin-Selenium-Sodium Pyruvate (ITS-A) (100X) (Cat. No. 51300044) are not animal origin-free.

Animal origin-free product versions are available for the ITS-G and ITS-X formulations: Insulin-Transferrin-Selenium (ITS-G) Select Supplement (100X) (Cat. No. A4000046501 and A4000046502) and Insulin-Transferrin-Selenium-Ethanolamine (ITS-X) Select Supplement (100X) (Cat. No. A4000046401 and Cat. No. A4000046402.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.