Geltrex™ Flex LDEV-Free hESC-Qualified Reduced Growth Factor Basement Membrane Matrix
Geltrex™ Flex LDEV-Free hESC-Qualified Reduced Growth Factor Basement Membrane Matrix
Geltrex™ Flex LDEV-Free hESC-Qualified Reduced Growth Factor Basement Membrane Matrix
Gibco™

Geltrex™ Flex LDEV-Free hESC-Qualified Reduced Growth Factor Basement Membrane Matrix

Geltrex Flex LDEV-Free hESC-Qualified Reduced Growth Factor Basement Membrane Matrix is a soluble basement membrane matrix purified from EHS tumors. Each lot is tested with iPSCs and hESC cells to ensure reliability and consistency in hESC/iPSC studies.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A40000468025 mL
A40000468011 mL
A400004680310 mL
Catalog number A4000046802
Price (USD)
249.00
Each
Quantity:
5 mL
Price (USD)
249.00
Each

Gibco Geltrex Flex LDEV-Free hESC-Qualified Reduced Growth Factor Basement Membrane Matrix is a soluble form of basement membrane that is purified from Engelbreth-Holm-Swarm (EHS) tumor cells. The major components of Geltrex Flex matrix include laminin, collagen IV, entactin, and heparan sulfate proteoglycan.

Routinely used for attachment and maintenance of human embryonic stem cells (hESCs) and induced pluripotent stem cells (iPSCs), every lot of hESC-qualified Geltrex Flex matrix is quality control tested using hESCs and iPSCs to help ensure support of stem cell culture. For a neural induction of hESC and iPSC protocol, see Induction of Neural Stem Cells from Human Pluripotent User Guide (Pub. No. MAN0008031).

Features of Geltrex Flex LDEV-Free hESC-Qualified Reduced Growth Factor Basement Membrane Matrix

  • Each lot tested with iPSCs and H9 hESCs for at least five passages to help ensure reliability and consistency in hESC/iPSC studies
  • Each lot tested for the absence of lactose dehydrogenase elevating virus (LDEV), mycoplasma, and a broad panel of viruses and microorganisms
  • Lots tested for endotoxin, maintenance of gel stability, and HUVEC tube formation
  • Growth factor–reduced to improve lot-to-lot consistency
  • Formulated without phenol red to minimize potential for estrogen-like effects
  • Provided in recyclable glass vials
  • 1-mL, 5-mL, and 10-mL sizes for maximum flexibility. Use only what you need. Reduce need for extensive aliquoting and unwanted freeze-thaws.

For the culture of primary cells, such as epithelial cells, endothelial cells, and smooth muscle cells, and angiogenesis assays, neurite outgrowth assays, or bile canaliculi formation assays with polarized hepatocytes, standard Geltrex Flex matrix can be used.

For the culture of tumoroids and other organoids, Geltrex Flex LDEV-Free Organoid-Qualified Reduced Growth Factor Basement Membrane Matrix is recommended. Each lot of organoid-qualified Geltrex Flex matrix is tested and supports the culture of mouse intestinal organoids, mouse liver ductal organoids, and mouse airway organoids, as well as expansion of mouse small intestinal organoids for more than five passages with typical organoid budding morphology.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeHuman induced pluripotent stem cells (iPSC), human embryonic stem cells (hESC), HUVEC, primary epithelial cells, neurons, endothelial cells, smooth muscle cells, hepatocytes.
Concentration9 to 13 mg/mL
Endotoxin Level≤ 4.5 EU/mL
Product TypeBasement Membrane Matrix
Purity or Quality GradeCell Culture Grade
Quantity5 mL
Shelf Life2 Years
Shipping ConditionDry Ice
SpeciesMouse
ClassificationReduced Growth Factor
Culture TypeAdherent Cell Culture, Mammalian Cell Culture, Stem Cell (Human, iPS - Induced Pluripotent Stem, Embryonic)
FormFrozen
Serum LevelSerum-free
SterilitySterile
Without AdditivesNo Phenol Red
Unit SizeEach

Frequently asked questions (FAQs)

Do you still recommend using Gibco Vitronectin as the matrix for Gibco Essential 8 Flex Medium?

Both Gibco Vitronectin (VTN-N) Recombinant Human Protein, Truncated (Cat. No. A400457 and A400458) and Gibco Geltrex Flex LDEV-Free hESC-qualified Reduced Growth Factor Basement Membrane Matrix (Cat. No. A4000046801, A4000046802, or A4000046803) have been successfully used with Essential 8 Flex Medium.

What substrate should I use for cardiomyocyte differentiation?

We recommend using Gibco Geltrex Flex LDEV-Free hESC-qualified Reduced Growth Factor Basement Membrane Matrix (Cat. No. A4000046801, A4000046802, or A4000046803) or Gibco Vitronectin (VTN-N) Recombinant Human Protein, Truncated (Cat. No. A400457 or A400458) for xeno-free applications.

You recommend Geltrex matrix as the matrix for routine culture of StemFlex Medium. Can I also use Vitronectin or rhLaminin-521?

Yes. We recommend following the coating instructions for Vitronectin or rhLaminin-521 and using the culture and passaging recommendations in the StemFlex protocol.

I am interested in using StemFlex Medium in somatic cell reprogramming. What protocol should I follow for human fibroblasts?

We have evaluated the use of Cytotune-iPS 2.0 Sendai Reprogramming Kit (Cat. No. A16517) for somatic cell reprogramming of both neonatal and adult human dermal fibroblasts. For fibroblasts, follow the instructions provided in the Cytotune 2.0 reprogramming manual for feeder-free reprogramming (pgs. 16-20). On Day 7, you may use rhVTN-N (Cat. No. A400457 or A400458), Geltrex Flex matrix (Cat. No. A4000046801, A4000046802, or A4000046803), or rhLaminin-521 (Cat. No. A29248 or A29249). From Day 8 onward, rather than feeding daily with Essential 8 Medium, we recommend that you feed reprogrammed fibroblasts every-other-day with StemFlex Medium.

I am interested in using StemFlex Medium in somatic cell reprogramming. What protocol should I follow for blood cells?

We have evaluated the use of Cytotune-iPS 2.0 Sendai Reprogramming Kit (Cat. No. A16517) for somatic cell reprogramming of CD34+ blood cells. For CD34+ cells, follow the instructions provided in the Cytotune 2.0 reprogramming manual for feeder-free reprogramming (pgs. 39-44). On Day 3, you can utilize rhVTN-N (Cat. No. A400457 or A400458), Geltrex Flex matrix (Cat. No. A4000046801, A4000046802, or A4000046803), or rhLaminin-521 (Cat. No. A29248 or A29249). From Day 8 onward, rather than feeding daily with Essential 8 Medium, reprogrammed CD34+ cells should be fed every-other-day with StemFlex Medium.

Citations & References (5)

Citations & References
Abstract
Alzheimer's disease pathology degrades an NMDA receptor-dependent spontaneous activity pattern in cortico-hippocampal circuits.
Authors:Ellingford R,Harris SS,Kehring M,Rajani RM,Lam FKW,Graykowski D,Bӧken D,Welikovitch LA,Khasnavis A,Laban R,Heslegrave A,Yaman U,Mate de Gerando A,Bond SA,Wray S,Salih DA,Dupret D,Dolan RJ,Klenerman D,Zetterberg H,Hyman BT,Busche MA
Journal:Neuron
PubMed ID:41916281
Memory-based cognition relies on the integrity of cortico-hippocampal circuits, which are compromised in Alzheimer’s disease (AD) as Aβ and tau accumulate. However, the mechanisms linking this pathology to circuit dysfunction remain unclear. In mouse models, using in vivo two-photon and Neuropixels recordings, we show that Aβ−tau pathology promotes both region- ... More
Epithelial cells fire voltage spikes.
Authors:Yu SM,Granick S
Journal:Proceedings of the National Academy of Sciences of the United States of America
PubMed ID:42301785
Bioelectric signaling is well characterized in neurons and cardiomyocytes but remains largely unexplored in epithelia. Using multielectrode arrays, we demonstrate that localized laser injury to epithelial monolayers (primary human keratinocytes and MDCK cells) triggers voltage spikes in the range of 4 to 12 per min for over 60 min postinjury. ... More
Are microcystin-LR and cylindrospermopsin neurotoxic for human cells? An in vitro study on induced pluripotent stem cells-derived models.
Authors:Casas-Rodríguez A,Pamies D,Lopez-Rodriguez D,Frangiamone M,Cameán AM,Zurich MG,Jos A
Journal:Toxicology
PubMed ID:42508561
Microcystin LR (MC LR) and cylindrospermopsin (CYN) are cyanobacterial toxins commonly detected during harmful algal blooms in freshwater systems. Their increasing occurrence raises concerns about water quality and potential risks to human health. Although the liver is their target organ, these cyanotoxins can also affect the nervous system. The potential ... More
Engineered human iPSC-derived dendritic cells dressed with tumor MHC complexes as a cancer vaccine.
Authors:Xu H,Kamei C,Gökbuget D,Du J,Barrero M,Wang L,Iwamoto Y,Soriano JM,Godding K,Wang B,Kwek SS,Nistor GI,Keirstead HS,Fong L,Kaneko S,Gaensler KML,Blelloch R
Journal:Cell stem cell
PubMed ID:42641609
Autologous-derived dendritic cells (DCs) are a promising source for cell-based cancer vaccines. However, their therapeutic potential is challenged by the number and quality produced and the diversity of antigens presented. To address these limitations, we present an approach that involves differentiating universal MHC-deficient human-induced pluripotent stem cells (hiPSCs) into CCR7 ... More
Neuron-derived SPP1 instructs microglia to limit degeneration.
Authors:Li S,Jakobs TC
Journal:Science advances
PubMed ID:42664340
Neurons actively shape immune responses that maintain central nervous system integrity. We identify SPP1 (secreted phosphoprotein 1) as a neuron-derived signal that reprograms microglia into a neuroprotective, homeostatic state after injury and during neurodegeneration. In mouse models of glaucoma and optic nerve damage, neuronal SPP1 enhances microglial autophagy, debris clearance, ... More