InnoviGene™ Suite Software
Applied Biosystems™

InnoviGene™ Suite Software

InnoviGene Suite software enables you to analyze and manage capillary electrophoresis sequencing & fragment analysis data generated by Applied Biosystems genetic analyzers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeLicense
A40001760Sequencing Software180 Days (Demo)
A40004750Fragment Analysis Software180 Days (Demo)
A40004747Fragment Analysis Software1 Year
A40004740Fragment Analysis Software3 Years
A40004752Fragment Analysis Software5 years
A40004743Fragment Analysis Software Renewal1 Year
A40004741Fragment Analysis Software Renewal3 Years
A40004739Fragment Analysis Software Renewal5 Years
A40004746Sanger Sequencing & Fragment Analysis Software180 Days (Demo)
A40004742Sanger Sequencing & Fragment Analysis Software1 Year
A40004744Sanger Sequencing & Fragment Analysis Software3 Years
A40004749Sanger Sequencing & Fragment Analysis Software5 years
A40004748Sanger Sequencing & Fragment Analysis Software Renewal1 Year
A40004751Sanger Sequencing & Fragment Analysis Software Renewal3 Years
A40004745Sanger Sequencing & Fragment Analysis Software Renewal5 Years
A40001762Sequencing Software1 Year
A40001765Sequencing Software3 Years
A40001763Sequencing Software5 Years
Catalog number A40001760
Price (USD)
-
Product Type:
Sequencing Software
License:
180 Days (Demo)

Applied Biosystems InnoviGene Suite software enables you to analyze and manage capillary electrophoresis sequencing and fragment analysis data generated by Applied Biosystems genetic analyzers. The software provides a streamlined workflow to guide you through uploading and analyzing the data and generating reports.

InnoviGene Suite includes SAE administrator console software with security, audit, and electronic signature (SAE) functions that support compliance with guidelines on electronic records security and prevents unauthorized access to the instrument.

The software is delivered via a download link sent to you after purchase.

Applications within InnoviGene Suite

InnoviGene Suite allows you to create projects for use with other applications that are integrated into the software. These applications enable you to complete an end-to-end workflow using projects that contain related files. The applications available depend on the type of InnoviGene Suite purchased ('sequencing' or 'fragment analysis').

Sequence Quality Check application (Innovigene Suite for sequencing)—enables you to perform the following activities:

  • View the quality information for trace files
  • Change quality settings and reanalyze trace files
  • Perform comparative analysis across multiple trace files

Sequence Identity application (InnoviGene Suite for sequencing)—enables you to view the assembly of multiple files compared to a reference and view basecall mismatches. Specifically, you can perform the following activities with the application:

  • Basecalling, quality value assignment, and mixed base identification
  • Trimming low-quality ends from each sequence, if the setting is active
  • Identifying poor-quality samples
  • Assembling the samples against the reference sequence and generating a specimen consensus sequence
  • Reviewing the basecalling quality values and consensus quality values
  • Identifying mismatches by aligning and comparing specimen sequences to the reference sequence. A mismatch is a position in the specimen that does not match with the reference sequence. Mismatches include base changes, insertions, and deletions.
  • Assigning specimen status based on alignment with the reference sequence and coverage criteria
  • Editing basecalls in samples or the consensus
  • Generating reports and results for export in CSV format

STR Typing application (InnoviGene Suite for fragment analysis)—the STR Typing application is genotyping software that uses short tandem repeat (STR) data. It enables you to perform the following activities:

  • Analyze a mixture of DNA fragments, separated by size
  • Provide a profile of the separation
  • Calculate the sizes of the fragments
  • Determine the alleles present in fragment analysis sample (FSA) files
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Sanger Sequencing
For Use With (Equipment)Applied Biosystems™ Genetic Analyzers
FormatSoftware
License180 Days (Demo)
Product TypeSequencing Software
Quantity5 Users
Sequencing TypeSanger Sequencing
Product LineInnoviGene
Unit SizeEach

Frequently asked questions (FAQs)

Can our existing SeqScape project templates or ROIs be used or adapted for the InnoviGene Suite?

SeqScape templates and RDGs cannot be imported into the InnoviGene Suite. If you have a GenBank file (.gb file) with non-overlapping ROIs, it can be imported into the InnoviGene Suite. Please note that some features in .gb files may be ignored by the InnoviGene Suite. For detailed information on .gb file compatibility, please refer to the relevant sections in the Getting Started Guide (GSG) or User Guide (UG) that can be found here: https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN1000299-InnoviGeneSuiteSoftware-EFI.pdf

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center and Capillary Electrophoresis Software Support Center.

Where is the data stored in InnoviGene Suite, and will it be accessible to other members of our group later?

The InnoviGene Suite data is stored on your local computer in a folder location specified by the user during the installation of InnoviGene Suite software. If another member of your group is added to the same SAE Administrator Console, the InnoviGene Suite allows you to share items with those same users within the InnvoiGene Suite. If they are using a different SAE Administrator Console License, trace files, projects, and other files can be exported and later imported into another InnoviGene Suite instance

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center and Capillary Electrophoresis Software Support Center.

.

Since the InnoviGene Suite is browser-based, can you confirm that our data is not being transmitted outside our computer systems (e.g., back to ThermoFisher Connect)?

The InnoviGene Suite does not have direct access to ThermoFisher Connect. With browser-based access, only colleagues on the same network and with user credentials on the same instance of the InnoviGene Suite may have access to the data. During installation of InnoviGene Suite software, the user will specify a folder location where all project data will be stored. Users will have to export files from the InnoviGene Suite and send to Thermo Fisher Scientific personnel, for troubleshooting purposes or data review.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center and Capillary Electrophoresis Software Support Center.

Can InnoviGene Suite software be co-installed with Applied Biosystems legacy software?

Co-installation with InnoviGene Suite software was tested for the following software:
- GeneMapper v6.1​
- Sequencing Analysis v8.0 (without the SDB license)
- SeqScape v4.1​
- Variant Reporter v3.1​
- Plate Manager v2.1.0

   Note: There is a known FlexNet licensing conflict between InnoviGene Suite and the SeqA 8 + SDB license and these softwares should not be co-installed.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center and Capillary Electrophoresis Software Support Center.

What antivirus software is the InnoviGene Suite compatible with?

The InnoviGene Suite is compatible with the following various antivirus software:
- Symantec™ Endpoint Protection
- McAfee®Endpoint Security
- Norton Internet Security™
- Microsoft® Defender antivirus software

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center and Capillary Electrophoresis Software Support Center.