CTS™ HBSS
Gibco™

CTS™ HBSS

Gibco Cell Therapy Systems (CTS) Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormat
A40002254032 LBag - BioProcess Container (BPC)
A40002254011 LBottle
Catalog number A4000225403
Price (USD)
385.00
Each
Quantity:
2 L
Format:
Bag - BioProcess Container (BPC)
Price (USD)
385.00
Each

Gibco Cell Therapy Systems (CTS) Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. This particular formulation lacks calcium, magnesium and phenol red.

Closed systems compatible
CTS HBSS is available in bottle and bioprocess container formats. The bioprocess container chamber is constructed from Aegis 5-14 film and includes sterile, weldable PVC/C-Flex hybrid tubing with a female Luer lock, providing versatile connection options for integrating into a closed system manufacturing process.

CTS quality
The Gibco CTS product line enables you to reduce your burden in qualifying reagents during your transition from research to clinical applications.

Products are manufactured at a site that uses methods and controls that conform to cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485- and ISO 9001–certified. Gibco CTS products follow USP <1043> ancillary materials for cell, gene, and tissue-engineered products within the responsibilities applicable to a supplier.

Gibco CTS products are of high quality and are supplied with harmonized documentation such as certificates of analysis and certificates of origin.

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
Specifications
Culture TypeMammalian and Stem Cell Culture
For Use With (Application)Cell and Gene Therapy Research, Development, and Manufacturing
FormatBag - BioProcess Container (BPC)
Manufacturing QualityISO13485, MDSAP, FDA-registered, 21CFR820
Quantity2 L
Shelf Life12 Months from Date of Manufacture
Shipping ConditionAmbient
FormClear, colorless solution
Product TypeHanks' Balanced Salt Solution
SterilitySterile-filtered
Sterilization Method0.2 μm Sterile-filtered
Unit SizeEach
Contents & Storage
15°C to 30°C