Zeba XL Spin Desalting Columns, 7K MWCO, 5 Columns - FAQs
Zeba XL Spin Desalting Columns, 7K MWCO, 5 Columns - FAQs
View additional product information for Zeba™ Spin Desalting Columns, Plates, and Cartridges, 7K MWCO, 0.5–100 mL - FAQs (89891, 89890, A40002469, 89808, A40002475, 89807, 89935, 89934, 89878, 89889, 89877, 89894, 89883, 89893, 89882, 89892)
3 product FAQs found
What reagents and conditions are Zeba Desalting columns, plates, and cartridges compatible with?
Zeba Desalting columns, plates, and cartridges are compatible with a range of reagents and conditions. They have been confirmed to be compatible with the following:
- pH range of 3-10
- Mild oxidants/reductants
- Chaotropes (stable in 8 M urea and 6 M guanidine HCl)
- Salts
- Alcohols up to 20%
- Organic solvents such as dimethyl sulfoxide (DMSO) and dimethylformamide (DMF) (we recommend using a step-gradient of increasing concentration of the organic solvent during column equilibration)
Find additional tips, troubleshooting help, and resources within our Protein Dialysis, Desalting, and Concentration Support Center.