CTS™ AIM V SFM Validation - FAQs
CTS™ AIM V SFM Validation - FAQs
View additional product information for CTS™ AIM V SFM Validation - FAQs (A4000311302)
4 product FAQs found
Will CTS AIM V SFM (Cat. No. 0870112BK) be discontinued?  
No. CTS AIM V SFM (Cat. No. 0870112BK) will not be discontinued. The same catalog number will be available with the new Aegis5-14 film following the planned transition in 2026. 
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Where can I get more information about Aegis 5-14 BPC (BioProcessing Container)? 
For additional information regarding Aegis 5-14 BPC, reach out to our Cell Culture Technical Support Team at techsupport@thermofisher.com. 
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What type of film is currently used for packaging CTS AIM V SFM (Cat. No. 0870112BK)?
CTS AIM V SFM (Cat. No. 0870112BK) is packaged in ASI 28 film, featuring an ethylene vinyl acetate (EVA) contact layer and four spike ports.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the difference between CTS AIM V SFM VALIDATION (Cat. No. A4000311302) and CTS AIM V SFM (Cat. No. 0870112BK)? 
The two media have the same formulation and GMP quality and are manufactured in ISO13485, MDSAP, FDA-registered, 21 CFR820 facilities. The main difference lies in the packaging. CTS AIM V SFM VALIDATION (Cat. No. A4000311302) is packaged in the Aegis 5-14 BioProcessing Container (BPC) with a low-density polyethylene (LDPE) contact layer and updated tubing and connector options. On the other hand, the current CTS AIM V SFM (Cat. No. 0870112BK) is packaged in ASI 28 film with an ethylene vinyl acetate (EVA) contact layer and four spike ports.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.