CTS™ AIM V MINUS Validation - FAQs
CTS™ AIM V MINUS Validation - FAQs
View additional product information for CTS™ AIM V MINUS Validation - FAQs (A4000311402)
4 product FAQs found
Will CTS AIM-V Medium without phenol red, without antibiotics (Cat. No. A3830802) be discontinued?
No. CTS AIM-V Medium without phenol red, without antibiotics (Cat. No. A3830802) will not be discontinued. The same catalog number will be available with the new Aegis5-14 film following the planned transition in 2026. 
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What type of film is currently used for packaging CTS AIM-V Medium without phenol red, without antibiotics (Cat. No. A3830802)?
CTS AIM-V Medium without phenol red, without antibiotics (Cat. No. A3830802) is packaged in ASI 28 film, featuring an ethylene vinyl acetate (EVA) contact layer and four spike ports.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the difference between CTS AIM V MINUS VALIDATION Medium (Cat. No. A4000311402) and CTS AIM-V Medium without phenol red, without antibiotics (Cat. No. A3830802)?
The two media have the same formulation and GMP quality and are manufactured in ISO13485, MDSAP, FDA-registered, 21 CFR820 facilities. The main difference lies in the packaging. CTS AIM V MINUS VALIDATION Medium (Cat. No. A4000311402) is packaged in the Aegis 5-14 BioProcessing Container (BPC) with a low-density polyethylene (LDPE) contact layer and updated tubing and connector options. On the other hand, CTS AIM-V Medium without phenol red, without antibiotics (Cat. No. A3830802) is packaged in ASI 28 film with an ethylene vinyl acetate (EVA) contact layer and four spike ports.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Where can I get more information about Aegis 5-14 BPC (BioProcessing Container)?
For additional information regarding Aegis 5-14 BPC, reach out to our Cell Culture Technical Support Team at techsupport@thermofisher.com. 
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.