KingFisher Compact 96 Deep Well Plates, 25 Pieces
Thermo Scientific™
KingFisher Compact 96 Deep Well Plates, 25 Pieces
The KingFisher™ Compact 96 Deep-well Plate is a single-use only consumable that processes up to 16 samples at the same time.
Have Questions?
Catalog NumberQuantity
A4000725325 Units/Box
Catalog number A40007253
Price (USD)
180.00
Each
Quantity:
25 Units/Box
Price (USD)
180.00
Each
The KingFisher™ Compact 96 Deep-well Plate is a single-use only consumable that processes up to 16 samples at the same time.
For research use only. Not intended for diagnostics procedures.
Specifications
DisposableSingle-use
Product TypeDeep-well Plate
Quantity25 Units/Box
Array8 x 12
BarcodeNo Barcode
ColorWhite
MaterialVirgin Polypropylene
No. of Wells96
SterilityNon-sterile
Surface TreatmentNon-treated
Well ShapeNot Applicable
Unit SizeEach