AmpQuest™ 96 Real-Time PCR System
Applied Biosystems™

AmpQuest™ 96 Real-Time PCR System

The AmpQuest 96 Real-Time PCR System is an affordable, high-performance real-time PCR instrument designed for research and routine testing. With a streamlined touchscreen interface and intuitive desktop software, the system pairs ease-of-use with quality and performance.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberExternal Computer
A40007908Not Included
A40007906Desktop
A40007907Laptop
Catalog number A40007908
Price (USD)
-
External Computer:
Not Included

The Applied Biosystems AmpQuest 96 Real-Time PCR System is an affordable, high-performance instrument designed for research and routine testing, addressing modern real-time PCR (qPCR) needs while preserving the core qualities of Applied Biosystems qPCR systems. Powered by Diomni Design & Analysis Software and featuring a streamlined touchscreen interface and reliable service and support, the AmpQuest 96 Real-Time PCR System enables data accuracy, ease of use, and versatility in applications.

Features of the AmpQuest 96 system include:

  • 96-well 0.2 mL format
  • Factory calibration and no recalibration at fixed time intervals required
  • OptiFlex technology (five coupled channels with white LED) for trusted results
  • VeriFlex block with six independent programmable temperature zones (25°C range)
  • Simplified instrument software with interactive touchscreen for greater ease of use
  • Diomni Design and Analysis Software v3 (desktop version)

Minimized cost of ownership

The AmpQuest 96 Real-Time PCR System is fully calibrated and ready to use upon installation. It supports the following common, popular fluorophores: FAM, SYBR Green, VIC, ABY, NED, TAMRA, JUN, ROX, Mustang Purple, and Cy5 dyes. Recalibration of the instrument at fixed time intervals is not required. Warranty extension and service contract options to fit any budget are available.

Multiplex capabilities minimize consumable/sample requirements

Multiplexing is a cost-effective solution to help simply and efficiently address the challenges of limited samples and expensive analysis. Five coupled excitation and emission filter channels, with white LED, enable up to 5-target multiplexing, supported by improved well-to-well and instrument-to-instrument data accuracy from OptiFlex technology.

Performance you can trust

Detect differences in target quantity as small as 1.5-fold, sensitivity down to one copy per reaction, and a linear dynamic range of 10 orders of magnitude.

Feature-rich software designed for research

Diomni Design and Analysis Software v3 (desktop version) is a feature-rich software solution that offers plate setup, quality checks, results review, and a multitude of single-plate application analysis modules.

High application versatility

Simple and optimized protocols, reliable assays and reagents, and intuitive software are available for a broad range of applications, including gene expression, genetic variation, animal health, and more.

System configuration flexibility

Option 1: Connect a co-located computer directly to the instrument and send experiments to and from the instrument. This connected computer can be procured along with the AmpQuest 96 system, or you can connect your own laptop or desktop.

Option 2: Network the instrument to allow remote access via a local area network (LAN).

Option 3: Operate in stand-alone mode (no connections) and transfer files via USB.

Options 2 and 3 do not require a co-located computer.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
CommunicationsEthernet, USB
Data AcquisitionWhole-plate imaging
Dimensions27 x 50 x 40 cm
Display TypeTouchscreen
Dynamic Range10 Logs
ExcitationBright white LED
External ComputerNot Included
Max. Ramp Rate6.5°C/sec.
Memory10 GB (2000 to 5000 run files)
MultiplexUp to 5 targets
Pre-Calibrated DyesYes
Precision1.5-fold
Quantity1 System
Reaction Volume Range10 to 100 μL
Run Time34 min.
Sample Ramp Rate3.66°C/sec.
Sensitivity1 Copy
Shipping ConditionRoom Temperature
Software CompatibilityDiomni™ Design & Analysis Software v3 (Desktop) or later
Thermal Accuracy±0.25°C
Thermal Range4°C to 99.9°C
Thermal Uniformity≤0.4°C Well-to-well
Voltage100/240 V ±10%
Weight<26 kg
Block Configurations96-well, 6-Zone VeriFlex Block
Block FormatFixed
Capacity96-well, 0.2 mL
Product TypeqPCR System
Unit SizeEach