RapidHIT™ ID ACE GlobalFiler™ Express Negative Control Cartridge
Applied Biosystems™
RapidHIT™ ID ACE GlobalFiler™ Express Negative Control Cartridge
The Applied Biosystems RapidHIT ID ACE GlobalFiler Express Negative Control Cartridge is for use with the Applied Biosystems RapidHIT ID System to verify instrument and/or reagent performance.
Have Questions?
Catalog NumberQuantity
A400080335 Pk.
Catalog number A40008033
Price (USD)
1,465.00
Each
Quantity:
5 Pk.
Price (USD)
1,465.00
Each
The Applied Biosystems RapidHIT ID ACE GlobalFiler Express Negative Control Cartridge is for use with the Applied Biosystems RapidHIT ID System to verify instrument and/or reagent performance. Each cartridge is individually wrapped and single-use/disposable.
Required for use:
  • Applied Biosystems RapidHIT ID System (Cat. No. A41810)
  • Applied Biosystems RapidHIT ID Instrument Software v1.3.1 or higher
  • Applied Biosystems RapidHIT ID Primary Cartridge (Cat. No. A53084)
For Criminal Justice and Forensic Use Only.
Specifications
For Use With (Application)Human Identification
For Use With (Equipment)RapidHIT™ ID System
Product TypeNegative Control
Quantity5 Pk.
FormatCartridge
Isolation TechnologyMembrane Filters
Sequencing TypeDNA and RNA Sequencing
Unit SizeEach