Genexus™ Conical Bottles
Ion Torrent™
Genexus™ Conical Bottles
Genexus Conical Bottles are installed in the sequencing reagents bay and serve as reservoirs for nucleotide reagent dilutions.
Have Questions?
Catalog NumberQuantity
A402751 Kit
Catalog number A40275
Price (USD)
858.00
Each
Quantity:
1 Kit
Price (USD)
858.00
Each
Genexus Conical Bottles are installed in the sequencing reagents bay of the Genexus Integrated Sequencer and serve as reservoirs for nucleotide reagent dilutions. They need to be replaced when filters in the bottles become partially clogged, causing decreased flow rate. If the sequencer detects restricted flow from the bottles during an instrument cleaning procedure, an alert will be displayed to replace the bottles.
For more information on when and how to replace the Genexus Conical Bottles, see the Genexus Integrated Sequencer User Guide.
For Research Use Only.
Specifications
For Use With (Equipment)Genexus™ Integrated Sequencer
Product TypeConical Bottles
Quantity1 Kit
Sequencing TypeNext Generation Sequencing
Unit SizeEach
Contents & Storage
1 Kit of 5 Conical Bottles
15°C to 30°C
Frequently asked questions (FAQs)
What is the level of detection for somatic variants with the Oncomine Myeloid Research Assay GX?
Detection of somatic variants with the Oncomine Myeloid Research Assay GX has been verified down to 5% allele frequency.
Which extraction kits are compatible with Oncomine Myeloid Research Assay GX?
Here is a list of kits that are compatible with Oncomine Myeloid Research Assay GX:
Nucleic acid isolation - RNA samples
- MagMAX mirVana Total RNA Isolation Kit, Cat. No. A27828
- PureLink Total RNA Blood Kit, Cat. No. K156001
- RNaseZap RNase Decontamination Solution, Cat. No. AM9780, AM9782, AM9784
Nucleic acid isolation - DNA samples
- MagMAX DNA Multi-Sample Ultra 2.0 Kit, Cat. No. A36570
- PureLink Genomic DNA Mini Kit, Cat. No. K1820-01
What are the sample input requirements for Oncomine Myeloid Research Assay GX?
The recommended minimum sample input is 27.75 ng of DNA (1.11 ng/µL minimum concentration) and 14.25 ng of RNA (0.95 ng/µL minimum concentration).
What sample controls should I use with the Oncomine Myeloid Research Assay GX?
The recommended controls are:
DNA Controls:
- Seraseq® Myeloid Mutation DNA Mix, Cat. No. 0710-0408 (SeraCare®)
- Myeloid DNA Reference Standard, Cat. No. HD829 (Horizon™)
- AcroMetrix™ Oncology Hotspot Control, Cat. No. 969056
RNA Controls:
- Universal Human Reference RNA, Cat. No. 740000 (Agilent™)
- Seraseq® Myeloid Fusion RNA Mix, Cat. No. 0710-0407 (SeraCare®)
How many Oncomine Myeloid Research Assay GX samples can be multiplexed on a GX5 Chip?
Multiplex sequencing of up to 8 Oncomine Myeloid Research Assay GX samples (8 DNA+ 8 RNA) per lane on a GX5 Chip can be performed in a single run. There are 4 lanes on the GX5 Chip, so 32 total samples (32 DNA+ 32 RNA) can be run on one GX5 Chip (8 samples per run).