MagMAX™ CORE Nucleic Acid Purification Kit + MagMAX™ CORE Mastitis & Panbacteria Module - FAQs
MagMAX™ CORE Nucleic Acid Purification Kit + MagMAX™ CORE Mastitis & Panbacteria Module - FAQs
View additional product information for MagMAX™ CORE Nucleic Acid Purification Kit + MagMAX™ CORE Mastitis & Panbacteria Module - FAQs (A40289)
4 product FAQs found
What kind of milk samples can be tested with the VetMAX MastiType kits?
All VetMAX MastiType kits may be used with milk, bulk milk, frozen milk, or preserved milk (e.g., with bronopol).
What qPCR instrument can I use to run the VetMAX MastiType kits?
Either the Applied Biosystems 7500/7500 Fast Real-Time PCR System or the QuantStudio 5 Real-Time PCR System may be used to run the VetMAX MastiType kits.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Which sample prep kit do you recommend upstream of the VetMAX MastiType kits?
We recommend using the MagMAX CORE Nucleic Acid Purification Kit + MagMAX CORE Mastitis & Panbacteria Module (Cat. No. A40289) with the VetMAX MastiType kits.
Which organisms can be detected by each VetMAX MastiType kit?
The VetMax MastiType Micro4 Kit detects Mycoplasma bovis, Staphylococcus aureus, Streptococcus agalactiae, and Streptococcus uberis.
The VetMax MastiType Myco8 Kit detects Mycoplasma spp., Mycoplasma alkalescens, Mycoplasma bovis, Mycoplasma bovigenitalium, Mycoplasma canadense, Mycoplasma californicum, Staphylococcus aureus, and Streptococcus agalactiae.
The VetMax MastiType Multi Kit detects Staphylococcus aureus, Staphylococcus spp. (including all major coagulase-negative staphylococci), Streptococcus agalactiae, Streptococcus dysgalactiae, Streptococcus uberis, Escherichia coli, Enterococcus spp. (incl. E. faecalis and E. faecium), Klebsiella oxytoca and/or Klebsiella pneumoniae, Serratia marcescens, Corynebacterium bovis, Trueperella pyogenes and/or Peptoniphilus indolicus, Staphylococcal beta-lactamase gene (penicillin-resistance gene), Mycoplasma bovis, Mycoplasma spp., Yeast, and Prototheca spp.