Product Image
Applied Biosystems™

QuantStudio™ 3 Real-Time PCR System, MiniAmp™ Plus Thermal Cycler, and 96-Well Plates Package

This package includes of a QuantStudio 3 qPCR system with laptop, MiniAmp Plus Thermal Cycler, and plastics consumables.QuantStudio 3 Real-TimeRead more
Have Questions?
Catalog NumberQuantity
A40393Package Solution
Catalog number A40393
Price (USD)
50,263.46
Each
Quantity:
Package Solution
Price (USD)
50,263.46
Each
This package includes of a QuantStudio 3 qPCR system with laptop, MiniAmp Plus Thermal Cycler, and plastics consumables.

QuantStudio 3 Real-Time PCR System
The Applied Biosystems QuantStudio 3 Real-Time PCR System is designed for users who need an affordable, easy-to-use real-time PCR system that doesn't compromise performance and quality. The simplified Design and Analysis software is ideal for both first-time and experienced users. When connected to the Thermo Fisher Cloud, the QuantStudio 3 system provides access to your data wherever and whenever you want. Using proven OptiFlex technology (featuring four coupled channels and white LED) and featuring three independent Veriflex temperature zones, the QuantStudio 3 system enables improved data accuracy and sensitivity for a broad range of genomic applications.

Features of the QuantStudio 3 system include:
• Simplified instrument software with interactive touchscreen for greater ease of use
• Proven OptiFlex technology (four coupled channels with white LED) for trusted results
• Secure cloud-based software option provides increased accessibility and storage options
• User account management, locked workflow, and Pause features for greater control of experiments
• Comes with SmartStart Orientation to get you up and running quickly.  Training in your laboratory includes basic PCR and qPCR fundamentals and setup with ThermoFisher Cloud.

Learn more about the QuantStudio 3 system >

MiniAmp Plus Thermal Cycler
The Applied Biosystems MiniAmp Plus Thermal Cycler is a compact, accurate, and economical thermal cycler designed to fit your lab’s basic thermal cycling needs. Innovations like a responsive color touch-screen and VeriFlex Blocks temperature-control technology enable simple, accurate optimization. The MiniAmp Plus Thermal Cycler can be accessed through the Thermo Fisher Cloud, giving you the freedom to design and securely upload your methods from any mobile device or desktop computer.

Key features and benefits:
• VeriFlex Blocks—with three independent temperature zones for precise temperature control over your PCR optimization
• 5-inch intuitive color touch-screen—for easy programming and training of new users
• Small footprint—With dimensions of 19 cm wide and 39 cm deep, you can fit the MiniAmp Thermal Cycler almost anywhere. And because its airflow is front to back, you can set multiple units side by side to save valuable bench space.
• WiFi-enabled and accessible by Thermo Fisher Connect—giving you the freedom to design and securely upload your methods from any mobile device or desktop computer

MicroAmp Optical 96-Well Reaction Plate with Barcode & Optical Adhesive Films
The Applied Biosystems MicroAmp Optical 96-Well Reaction Plate is optimized to provide unmatched temperature accuracy and uniformity for fast, efficient PCR amplification. This plate, constructed from a single rigid piece of polypropylene in a 96-well format, is compatible with Applied Biosystems 96-well real-time PCR systems and thermal cyclers.
• Frosted to minimize interfering fluorescence from wells of cycling block
• Screened to eliminate auto-fluorescent plates
• Unique design provides a barrier to ambient air to help insure well-to-well temperature uniformity
• Each reaction plate includes a unique serialized, eight-character number label (barcode) that is user-readable and machine-readable to prevent tracking errors

The included MicroAmp Optical Adhesive Films reduce the chance of well-to-well contamination and sample evaporation when applied to the microplates.
• Pressure-sensitive adhesive provides a tight seal over every well
• Will not interfere with sample reading

• Confirm correct alignment to plate, then press to seal• Easy to apply; does not stick to your glove
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Display TypeTouchscreen
For Use With (Equipment)QuantStudio™
Format96-well Plate
High-throughput CompatibilityHigh-throughput Compatible
Product LineApplied Biosystems
QuantityPackage Solution
Reaction SpeedStandard
Shipping ConditionApproved for Shipment at Room Temperature or on Wet Ice
Capacity96-well x 0.2 ml
Unit SizeEach
Package Components
MiniAmp™ Plus Thermal Cycler
Catalog Number
A37835
Unit Size
1 instrument
Qty
1
SMARTSTART ORNTATION,HALFDAY
Catalog Number
A28806
Unit Size
1 each
Qty
1
Contents & Storage
QuantStudio 3 Real Time PCR System, 96-well, 0.2 mL, Laptop, MiniAmp Plus Thermal Cycler, MicroAmp Optical 96-well Plates w/Barcode, Optical Adhesive Films (100 Plates and 100 Seals)

Frequently asked questions (FAQs)

What is the difference between continuous melt curves and step and hold melt curves on the Applied Biosystems Real-Time PCR Instruments?

Continuous melt curve: increases the temperature by the ramp increment (degrees C/sec) so that the temperature will change at a constant rate set in the protocol.
Step and Hold melt curve: increases the temperature by the ramp increment (degrees C) and then holds at that temperature for the time specified by the user.

The amount of data collected will depend on the filter(s) selected. The more filters you use, the longer the acquisition will take and the fewer data points will be collected.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Can I downgrade the firmware on my QuantStudio 5?

You can perform this action if you have access to an Admin account on the QuantStudio instrument and if it is compatible for the downgrade (newer instruments are not compatible with v1.3.3). If your instrument serial number is below the following, then it can be downgraded:
QuantStudio 5 96-well, 0.1 mL block - SN 272514140
QuantStudio 5 96-well, 0.2 mL block - SN 2725210507
QuantStudio 5 384 well block - SN 272533285
Please also check if the instrument is being used with any validated protocols that may require a particular firmware version to be compliant. If you need the firmware itself, you can download it here.
The instructions to downgrade the firmware are on p.4 of the Release Notes: QuantStudioTM 3 & 5 Touchscreen Instrument Operating Software Version 1.3.3. It's the same process as an upgrade.

What is the admin pin for QuantStudio 3 and 5 Real-Time PCR Systems?

If the admin PIN has not been changed, 1234 should work. If it does not work, you can also try 0000.
However, if the admin PIN has been changed and none of the above PINs work, follow the instructions in the QuantStudio 3 and 5 Real-Time PCR Systems manual, page 100.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Can I import a standard curve for my real-time PCR data into the Design & Analysis Software (v2.x)?

It is possible to import a standard curve into the Design & Analysis Software (v2.x) if you are analyzing results from one of the following Applied Biosystems real-time PCR instruments:
• 7900HT Fast Real-Time PCR System
• 7500/7500 Fast Real-Time PCR System
• StepOne/StepOne Plus Real-Time PCR System
• ViiA 7 Real Time PCR System
• QuantStudio 1 Real-Time PCR System
• QuantStudio 3 & 5 Real-Time PCR System
• QuantStudio 6 & 7 Flex Real-Time PCR System
• QuantStudio 6 & 7 Pro Real-Time PCR Systems
• QuantStudio 12K Flex Real-Time PCR system

Follow the instructions below to import a standard curve for analysis in the Design & Analysis Software (v2.x):
1. Open the data file in the Design & Analysis Software (v2.x).
2. Click on Actions.
3. Click on "Standard Curve Analysis Setting".
4. Select "External Standard Curves" and click on "Import".
5. Browse to the proper .csv file and click "Open".
6. Click "Apply".

Can I import a standard curve for my real-time PCR data into the QuantStudio Design & Analysis Software (v1.x)?

If you are analyzing results from the QuantStudio 1 Real-Time PCR System, QuantStudio 3 Real-Time PCR System, or QuantStudio 5 Real-Time PCR System, it is possible to import a standard curve into the QuantStudio Design & Analysis Software (v1.x) following the instructions below:
1. Open the data file in the QuantStudio Design & Analysis Software (v1.x).
2. Go to Analysis Settings.
3. Click on the "Standard Curve Settings" tab.
4. Select the option to "Use a standard curve imported from another experiment" and click on "Import".
5. Browse to and highlight the proper data file and click "Select".
6. Click Apply.