Human PRKCD (PKC delta), GST Tag Recombinant Protein - FAQs
Human PRKCD (PKC delta), GST Tag Recombinant Protein - FAQs
View additional product information for Human PRKCD (PKC delta), GST Tag Recombinant Protein - FAQs (A41912, A41913, A41911)
1 product FAQs found
Which kinase, similar to PCK, would be suitable for protein phosphorylationprotein?
The PRKCD (PKC delta) Recombinant Human Protein (Cat. No. A41913). This recombinant protein with a GST Tag produced on insect cells has been validated for Kinase assays thus it would be able to phosphorylate proteins.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.