MicroSEQ™ ID Microbial Identification Software, version 3.1.3 - FAQs
MicroSEQ™ ID Microbial Identification Software, version 3.1.3 - FAQs
View additional product information for MicroSEQ™ ID Microbial Identification Software, version 3.1.3 - FAQs (A41949)
3 product FAQs found
What computers are compatible with MicroSEQ ID Software v3.1.3 (system installation)?
The computer that is compatible with MicroSEQ ID Software v3.1.3 (system installation) is the Dell Optiplex XE2 with Win10 IOT 2016-64-bit.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What are the minimum system requirements for MicroSEQ ID Microbial Identification Software v3.1.3?
The minimum system requirements for MicroSEQ ID Microbial Identification Software v3.1.3 are the following:
- Intel Core i7 Processor, 3.1 GHz or faster
- Microsoft Windows 10 OS, Service Pack 1, 64-bit
- At least 16 GB of RAM
- At least 250 GB of disk space for software
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What is included in the MicroSEQ ID Microbial Identification Software v3.1.3, with All Software Libraries Kit (Cat. No. A48593)?
The MicroSEQ ID Microbial Identification Software v3.1.3, with All Software Libraries Kit (Cat. No. A48593) includes the following components:
MicroSEQ ID Microbial Identification Software v3.1.3
MicroSEQ ID Microbial Identification Software v3.1.3 License
MicroSEQ ID Supplemental Library v2019
MicroSEQ ID Supplemental Library v2019 License
MicroSEQ ID 16S rDNA 500 Microbial Library, v2019
MicroSEQ ID 16S rDNA 500 Microbial Library, v2019 License
MicroSEQ ID Fungal Gene Library, v2018
MicroSEQ ID Fungal Gene Library, v2018 License
MicroSEQ ID v2.0 Full Gene Library
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.