MicroSEQ™ ID 16S rDNA 500 Microbial Library, v2019 - FAQs
MicroSEQ™ ID 16S rDNA 500 Microbial Library, v2019 - FAQs
View additional product information for MicroSEQ™ ID 16S rDNA 500 Microbial Library, v2019 - FAQs (A41950)
3 product FAQs found
What is included in the MicroSEQ ID Microbial Identification Software v3.1.3, Lite, with All Software Libraries Kit (Cat. No. A48594)?
The MicroSEQ ID Microbial Identification Software v3.1.3, Lite, with All Software Libraries Kit (Cat. No. A48594) includes the following components:
MicroSEQ ID Microbial Identification Software, v3.1.3, Lite, WIN 10
MicroSEQ ID Microbial Identification Software, v3.1.3, Lite License
MicroSEQ ID Supplemental Library v2019
MicroSEQ ID Supplemental Library v2019 License
MicroSEQ ID 16S rDNA 500 Microbial Library, v2019
MicroSEQ ID 16S rDNA 500 Microbial Library, v2019 License
MicroSEQ ID Fungal Gene Library, v2018
MicroSEQ ID Fungal Gene Library, v2018 License
MicroSEQ ID v2.0 Full Gene Library
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What is included in the MicroSEQ ID Microbial Identification Software v3.1.3, with All Software Libraries Kit (Cat. No. A48593)?
The MicroSEQ ID Microbial Identification Software v3.1.3, with All Software Libraries Kit (Cat. No. A48593) includes the following components:
MicroSEQ ID Microbial Identification Software v3.1.3
MicroSEQ ID Microbial Identification Software v3.1.3 License
MicroSEQ ID Supplemental Library v2019
MicroSEQ ID Supplemental Library v2019 License
MicroSEQ ID 16S rDNA 500 Microbial Library, v2019
MicroSEQ ID 16S rDNA 500 Microbial Library, v2019 License
MicroSEQ ID Fungal Gene Library, v2018
MicroSEQ ID Fungal Gene Library, v2018 License
MicroSEQ ID v2.0 Full Gene Library
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What is included in the MicroSEQ ID Microbial Identification Software v3.1.3, Lite, with Software Library Bundle (Cat. No. A46530)?
The components of the MicroSEQ ID Microbial Identification Software v3.1.3, Lite, with Software Library Bundle (Cat. No. A46530) are the following:
- MicroSEQ ID Microbial Identification Software, v3.1.3, Lite, WIN 10
- MicroSEQ ID Microbial Identification Software, v3.1.3, Lite License
- MicroSEQ ID Fungal Gene Library, v2018
- MicroSEQ ID Fungal Gene Library, v2018 License
- MicroSEQ ID 16S rDNA 500 Microbial Library, v2019
- MicroSEQ ID 16S rDNA 500 Microbial Library, v2019 License
- MicroSEQ ID 16S rDNA Full Gene Library
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.