How many beads are in each tube of the MagMAX Microbiome Bead Tubes?
The number of beads in each tube is proprietary.
Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.
Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.