MagMAX™ Viral/Pathogen Wash Solution - FAQs
View additional product information for MagMAX™ Viral/Pathogen Wash Solution - FAQs (A42360)
1 product FAQs found
I am seeing some small particles floating in the MagMAX Viral/Pathogen Wash Solution. Does this affect the performance of the wash solution?
The MagMAX Viral/Pathogen Wash Solution may develop inert white or brown particulates that float in solution. This is not a cause for concern and does not negatively affect performance. This statement is also listed on the product page: https://www.thermofisher.com/order/catalog/product/A42360