MagMAX™ Viral/Pathogen Binding Beads
Applied Biosystems™

MagMAX™ Viral/Pathogen Binding Beads

These binding beads are included in the MagMAX Viral/Pathogen Nucleic Acid Isolation kits and MagMAX Microbiome Ultra Nucleic Acid IsolationRead more
Have Questions?
Catalog NumberQuantity
A4236220 mL
Catalog number A42362
Price (USD)
1,031.65
Online exclusive
1,146.00
Save 114.35
Each
Quantity:
20 mL
Price (USD)
1,031.65
Online exclusive
1,146.00
Save 114.35
Each
These binding beads are included in the MagMAX Viral/Pathogen Nucleic Acid Isolation kits and MagMAX Microbiome Ultra Nucleic Acid Isolation Kit (Cat. Nos. A42352, A42356, A42357, and A42358). They are made available separately for applications that require more binding beads than are provided in the kit.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
High-throughput CompatibilityHigh-throughput Compatible, Manual Protocols, Automated Protocols
Product LineMagMAX
Product TypeViral/Pathogen Binding Beads
Purification TargetViral DNA/RNA, Pathogen DNA/RNA, Total Nucleic Acids
Quantity20 mL
Sample TypeLiquid Samples (e.g. Serum), Viral Samples, Buccal Samples, Blood, Cells, Bacteria, Fungi, Yeast
Shipping ConditionRoom Temperature
For Use With (Application)Next-Generation Sequencing (NGS), Real Time PCR (qPCR), RT-PCR
Unit SizeEach
Contents & Storage
Store at room temperature.