Attune™ NxT and CytPix Maintenance Kit - FAQs
Attune™ NxT and CytPix Maintenance Kit - FAQs
View additional product information for Attune™ NxT and CytPix Maintenance Kit - FAQs (A43038)
3 product FAQs found
How often should I change filters for the Attune NxT Acoustic Focusing Cytometer?
Filters are recommended for replacement once every three months, unless performing small particle detection when once-a-month replacement is recommended.
Can I purchase the individual components of the Attune NxT Maintenance Kit as standalone products?
Yes. Here are the catalog numbers for the individual components:
- Cat. No. 100022156: 1 Waste bottle
- Cat. No. 100022155: 1 Focusing fluid bottle
- Cat. No. 100022587: 12 Focusing fluid filters
- Cat. No. 100022591: 2 Syringes
- Cat. No. 4490099: 1 SIP
- Cat. No. A10496: Debubble solution
Is the debubble solution provided in the Attune NxT Maintenance Kit the same as the standalone product, Attune Debubble Solution (1X), Cat. No. A10496?
Yes. It is the same 50 mL bottle of 1X Debubble Solution as provided in Attune Debubble Solution (1X).