LV-MAX™ Lentiviral Packaging Mix - FAQs
LV-MAX™ Lentiviral Packaging Mix - FAQs
View additional product information for LV-MAX™ Lentiviral Packaging Mix - FAQs (A43237)
3 product FAQs found
Are the LV-MAX Lentiviral Packaging Mix (Cat. No. A43237) and ViraPower Lentiviral Packaging Mix (Cat. No. K497500) interchangeable?
Yes, they can be used interchangeably.
Find additional tips, troubleshooting help, and resources within our Transfection Support Center.
What are the major differences between LV-MAX and CTS LV- MAX products?
The LV-MAX and CTS LV- MAX products have equivalent performance. In contrast to the research use only LV-MAX products, CTS LV-MAX products have an additional intended use for cell and gene therapy applications, and are complemented with a more robust regulatory support package to enable clinical therapeutic applications. This means that to the best of our ability, we keep our CTS product specifications and release requirements up-to-date in accordance with the latest regulatory guidance specific for this intended use.
What is the intended use claim for your LV-MAX and CTS LV-MAX products?
Our LV-MAX products are intended for research use only while our CTS LV- MAX products are intended for research use or manufacture of cell, gene or tissue based products. Please refer to the individual products for further details on the intended use claim.