Ion 550™ Kit for Agrigenomics—Chef - FAQs
Ion 550™ Kit for Agrigenomics—Chef - FAQs
View additional product information for Ion 550™ Kit for Agrigenomics—Chef - FAQs (A43542)
8 product FAQs found
Is Ion S5 Sequencing Reagents (Part No. A27768) used for both the Ion 540 and Ion 550 Chip sequencing?
Yes, Ion S5 Sequencing Reagents (Part No. A27768) is used on the instrument for both Ion 540 and Ion 550 Chip sequencing.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center
When setting up a single chip run with the Ion 550 Kit-Chef, I get an error message indicating that there is a chip type mismatch. What does it mean?
In most cases, a chip type mismatch error indicates that the Ion Chef S5 Series Chip Balance was placed incorrectly in the Chip-loading centrifuge. Please make sure that the Ion Chef S5 Series Chip Balance is placed in Position 2 of the Chip-loading centrifuge.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How do I ensure cartridge-use tracking with the Ion 550 Kit-Chef?
To use the flexible workflow feature (single chip/dual chip) and enable cartridge-use tracking between Torrent Servers across multiple Ion Chef instruments, Torrent Servers must be linked in an Ion Mesh configuration. For further information on how to link Torrent Servers in an Ion Mesh setup, go to Software Administration -> Set up Ion Mesh in Torrent Suite Software Help.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What version of Torrent Suite Software is compatible with the Ion 550 Kit-Chef?
The Ion 550 Kit-Chef is compatible with Torrent Suite Software version 5.8 and later.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What is the maximum library read length allowed with the Ion 550 Kit-Chef?
The Ion 550 Kit-Chef can be used with up to 200-base-read libraries of any type, prepared using any available Ion library kit.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
On which instrument platforms is the Ion 550 Kit-Chef supported?
The Ion 550 Kit-Chef is supported on the Ion GeneStudio S5 Plus System and Ion GeneStudio S5 Prime System. The Ion 550 Kit-Chef is also compatible with the Ion S5 XL System, but is not compatible with the Ion S5 System, nor the base model Ion GeneStudio S5 System.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How many chips can I run with the Ion 550 Kit-Chef?
The Ion 550 Kit-Chef includes reagents and materials that are required to use the Ion Chef System to prepare template-positive Ion Sphere particles from up to 200-base-read libraries and load eight Ion 550 chips (two chips/run). The kit also includes reagents and materials to sequence eight loaded chips (two sequencing runs/initialization).
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Can I reuse the Ion 550 Chef Reagents and Ion 550 Chef Solutions cartridges for running a second chip separately from the first chip?
The Ion 550 Chef Reagents and Ion 550 Chef Solutions cartridges can be reused in a second single-chip templating run if the second run is performed within eight days of the first run.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.