Product Image
Ion Torrent™

Ion 550™ Chip Kit for Agrigenomics

The Ion 550 Chip Kit contains chips for sample tracking and sequencing with the Ion GeneStudio S5 Plus, the IonRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A435624 Chips
A429628 Chips
Catalog number A43562
Price (MXN)
-
Quantity:
4 Chips
The Ion 550 Chip Kit contains chips for sample tracking and sequencing with the Ion GeneStudio S5 Plus, the Ion GeneStudio S5 Prime, and Ion S5 XL sequencing systems. The Ion 550 Chip generates 100–130 million sequencing reads by electronically detecting polymerase-driven base incorporation without the use of fluorescence. By eliminating the use of an optical detection system, this advance in next-generation sequencing technology allows for rapid sequencing times of as little as 2.5 hours for 200 bp sequencing.

The Ion 550 Chip Kit offers:
• Support for a variety of research applications, including sequencing of up to four human exomes, cell-free nucleic acid applications, transcriptome profiling, and targeted resequencing of gene panels
• A rapid run-time of as little as 2.5 hours for 200 bp sequencing
• Sample tracking functionality from an integrated barcode
Notes:The Ion 550 Chip is only compatible with the Ion GeneStudio S5 Plus, Ion GeneStudio S5 Prime, and Ion S5 XL systems.
Use this chip with the Ion 550 Kit—Chef for automated template preparation and chip loading.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Ion GeneStudio S5 Plus, Ion GeneStudio S5 Prime, and Ion S5 XL System
Product TypeChip Kit
Quantity4 Chips
Shipping ConditionRoom Temperature
Product LineIon Torrent
Unit SizeEach
Contents & Storage
Store at room temperature.

Frequently asked questions (FAQs)

How long can my loaded Ion 550 chip be stored prior to running on the sequencer?

Remove and sequence chips as soon as possible after the Ion Chef System finishes loading them. If you cannot sequence a loaded chip immediately, store it in a chip storage container at 4 degrees C until you are ready to run it (up to 24 hours maximum).

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

Why do my Ion 550 chip "Reagent Check" traces look different from those seen with the other Ion chips (510, 520, 530, and 540)?

This is because the new chip surface and the way the instrument reads the Ion 550 chip are different from the Ion 540/530/520/510 chips.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

How many Oncomine Comprehensive Assay v3 libraries may be pooled on a single Ion 550 chip?

The Oncomine Comprehensive Assay v3 is validated with 32 libraries per Ion 550 chip (16 samples DNA and RNA).

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

How many Ion AmpliSeq Human Exome libraries can be pooled on a single Ion 550 chip?

The Ion 550 workflow supports 3-4 Ion AmpliSeq Human Exome libraries per run.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

What are the shipping and storage conditions for the Ion 550 chip?

The Ion 550 chip is shipped at room temperature and should be stored at room temperature.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.