Pierce™ Pyrogen-Free Glass Test Tubes - FAQs
Pierce™ Pyrogen-Free Glass Test Tubes - FAQs
View additional product information for Pierce™ Pyrogen-Free Glass Test Tubes - FAQs (A43889)
1 product FAQs found
What are the dimension of the Pierce Pyrogen-Free Glass Test Tubes (Cat. No. A43889)?
The Pierce Pyrogen-Free Glass Test Tubes have an outer diameter of 13 mm and a length of 100 mm.