TaqMan™ Fast Advanced Master Mix for qPCR, no UNG, 1 mL - FAQs
TaqMan™ Fast Advanced Master Mix for qPCR, no UNG, 1 mL - FAQs
View additional product information for TaqMan™ Fast Advanced Master Mix for qPCR, no UNG - FAQs (A44359, A44360)
1 product FAQs found
I am using the TaqMan Gene Expression Cells-to-CT kit. Can I use the TaqMan Fast Advanced Master Mix in place of the TaqMan Gene Expression Master Mix to set up the qPCR reaction?
Yes, the TaqMan Fast Advanced Master Mix can be used in place of the TaqMan Gene Expression Master Mix when setting up the qPCR reaction for the TaqMan Gene Expression Cells-to-CT kit.