Ion GeneStudio™ S5 Plus Food Protection System
Thermo Scientific™

Ion GeneStudio™ S5 Plus Food Protection System

Identify meat, fish or plant species present in all food types using the Ion Genestudio S5 Plus Food Protection System.
Have Questions?
Número de catálogoCantidad
A446481 System
Número de catálogo A44648
Precio (MXN)
-
Cantidad:
1 System

Identify meat, fish or plant species present in all food types using the Ion Genestudio S5 Plus Food Protection System, a semiconductor-based next-generation sequencing (NGS) system which, when used as part of the Thermo Scientific™ Next Generation Sequencing Food Authenticity Workflow, allows identification of the species present in even the most challenging sample types.

The simple-to-use Ion GeneStudio S5 Prime Food Protection System is optimized to provide maximum throughput with minimum time-to-results for laboratories who need to accelerate their sequencing capabilities.

Identification of the species present in food and feed samples is a critical step for proof of authenticity, supply chain integrity, and quality control of handling and cleaning processes in food production lines.

The Thermo Scientific NGS Food Authenticity Workflow utilises Ion Torrent™ Next Generation Sequencing technology to enable an untargeted screening approach making it possible to identify the species contained in a food sample by comparison with a DNA database of meat, plant or fish species.

The Ion GeneStudio S5 Food Protection Series Sequencers are an integral component of the workflow. With cartridge-based reagents, the systems are simple to use and offer scalability and flexibility.

Key features Ion GeneStudio S5 Plus include:

• Easy-to-use with minimal hands on time: cartridge-based plug-and-play reagents
• Rapid data processing: complete sequencing run and analysis in hours
• Flexible throughput: select the chip appropriate to your throughput requirements (Ion S5 510–550 chip)
• Scalability: run all your targeted sequencing or microbial applications on a single sequencer. Scale your throughput using the range of Ion S5 Series chips depending on your application.
• Complete with laptop: suitable for use with SGS ALL Species ID Software


The Ion GeneStudio S5 Food Protection Series

The Ion GeneStudio sequencers share the same foundational technology and fluidic architecture across all three instruments, the Ion GeneStudio S5 Food Protection System, the Ion GeneStudio S5 Plus Food Protection System and the Ion GeneStudio S5 Prime Food Protection System. The Primary difference across the three instruments is on-sequencer analysis time.

The Ion GeneStudio S5 Food Protection Systems leverage the speed of semiconductor sequencing to enable the production of high-quality sequencing data in a few hours and enable you to go from DNA library to data in as little as 24 hours with only 45 minutes of total hands-on time, when paired with the Ion Chef™ Food Protection Instrument.

With single-day installation, a straightforward user interface, and cartridge-based reagents, the Ion GeneStudio S5 Food Protection Systems are simple and easy to use.

 

 Ion GeneStudio S5 Food Protection SystemIon GeneStudio S5 Plus Food Protection SystemIon GeneStudio S5 Prime Food Protection System
Max. throughput/day (chip type)15 Gb (one Ion 540 Chip)30 Gb (two Ion 540 chips)50 Gb (two Ion 550 chips)
Total sequencing and analysis time at max. throughput19 hr (Ion 540 Chip)10 hr (Ion 540 Chip)
11.5 hr (Ion 550 Chip)
8.5 hr (Ion 540 Chip)
8.5 hr (Ion 550 Chip)
Compatible chipsIon 510, 520, 530, 540 chipsIon 510, 520, 530, 540, and 550 chips
Para uso exclusivo en investigación. No debe utilizarse en los procedimientos de diagnóstico.
For Research Use Only. Not for use in diagnostic procedures.
Especificaciones
DescripciónIon GeneStudio S5 Plus Food protection system
Dimensiones exteriores (L × An × Al)54.2 x 80.6 x 50.9 cm
Requisitos eléctricos100-240 VAC, 50/60 Hz, 6.5-14.5 A
Para utilizar con (equipo)Ion 510 Chip, Ion 520 Chip, Ion 530 Chip, Ion 540 Chip
Altura exterior (imperial)20 in.
Altura exterior (métrico)50.9 cm
Longitud (imperial) exterior21.33 in.
Longitud (métrico) exterior54.2 cm
Entorno operativoTemperature 15-30 C, Humidity: 10-80%, Altitude: Upto 2,500 m (8,200ft) above sea level, Thermal output at typical power draw of 1,200 W: 4,094 BTU/hr
Cantidad1 System
Peso63.5 kg
Anchura exterior (imperial)31.73 in.
Anchura exterior (métrico)80.6 cm
Software TypeSGS All Species ID Software
Material de partidaADN
Unit SizeEach
Contenido y almacenamiento
Room temperature