2 M Citric Acid
Gibco™

2 M Citric Acid

Gibco concentrates are intended for dilution at the point of use and come sterile-filtered and packaged in conveniently sized bioprocess containers. Sourcing concentrated stock solutions can help streamline buffer preparation by helping reduce preparation time, storage space, and the manufacturing footprint required for hydration.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A44667BE100 L
A44667BA5 L
A44667BC20 L
A44667BD50 L
A44667BF200 L
Catalog number A44667BE
Price (USD)
-
Quantity:
100 L

Gibco concentrates are intended for dilution at the point of use and come sterile-filtered and packaged in conveniently sized bioprocess containers. Sourcing concentrated stock solutions can help streamline buffer preparation by helping reduce preparation time, storage space, and the manufacturing footprint required for hydration.

Features

  • Concentrated for dilution on-site
  • Available in a range of pack sizes
  • Established shelf life: set specifications
  • Made to order
  • Manufactured to ISO and cGMP standards to meet the stringent expectations of highly regulated industries
  • Readily accessible documentation to help accelerate qualification

Benefits

  • Helps ease time requirements for in-house buffer preparation
  • Mitigates space constraints for equipment storage
  • Streamlines in-house processes related to formulation, hydration, testing, and validation
  • Addresses workflow needs for biologics development and manufacturing stages

Additional information

  • Multi-compendial testing: compendial-grade or equivalent raw materials
  • Primary packaging: TAegis 5-14 single-use bioprocess container (BPC)
  • Secondary packaging: rigid shipping container with appropriate dunnage
  • Manufacturing facilities: ISO 13485–certified, cGMP-compliant to 21 CFR Part 820 standard
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
ColorClear
Concentration2 M
FormulationAcid/Buffer Component
Manufacturing QualityCGMP
Quantity100 L
Shelf Life12 Months
FormClear, colorless solution
Product TypeCitric Acid
SterilitySterile-filtered
Sterilization MethodSterile-filtered
pH0.8 to 1.2
Unit SizeEach