0.3 M Sodium Phosphate Dibasic
Gibco™

0.3 M Sodium Phosphate Dibasic

Gibco concentrates are intended for dilution at the point of use and come sterile-filtered and packaged in conveniently sized bioprocess containers. Sourcing concentrated stock solutions can help streamline buffer preparation by helping reduce preparation time, storage space, and the manufacturing footprint required for hydration.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
A44668BA5 L
A44668BC20 L
A44668BD50 L
A44668BE100 L
A44668BF200 L
Número de catálogo A44668BA
Precio (MXN)
-
Cantidad:
5 L

Gibco concentrates are intended for dilution at the point of use and come sterile-filtered and packaged in conveniently sized bioprocess containers. Sourcing concentrated stock solutions can help streamline buffer preparation by helping reduce preparation time, storage space, and the manufacturing footprint required for hydration.

Features

  • Concentrated for dilution on-site
  • Available in a range of pack sizes
  • Established shelf life: set specifications
  • Made to order
  • Manufactured to ISO and cGMP standards to meet the stringent expectations of highly regulated industries
  • Readily accessible documentation to help accelerate qualification

Benefits

  • Helps ease time requirements for in-house buffer preparation
  • Mitigates space constraints for equipment storage
  • Streamlines in-house processes related to formulation, hydration, testing, and validation
  • Addresses workflow needs for biologics development and manufacturing stages

Additional information

  • Multi-compendial testing: compendial-grade or equivalent raw materials
  • Primary packaging: TAegis 5-14 single-use bioprocess container (BPC)
  • Secondary packaging: rigid shipping container with appropriate dunnage
  • Manufacturing facilities: ISO 13485–certified, cGMP-compliant to 21 CFR Part 820 standard
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
ColorClear
Concentración0.3 M
FormulaciónPhosphate Buffer Component
Calidad de fabricaciónCGMP
Cantidad5 L
Duración de almacenamiento12 Months
FormularioClear, colorless solution
Tipo de productoSodium Phosphate Dibasic
EsterilidadSterile-filtered
Sterilization MethodSterile-filtered
pH8.8 to 9.1
Unit SizeEach