Sodium Phosphate Monobasic
Gibco™
Sodium Phosphate Monobasic
Gibco concentrates are intended for dilution at the point of use and come sterile-filtered and packaged in conveniently sized bioprocess containers. Sourcing concentrated stock solutions can help streamline buffer preparation by helping reduce preparation time, storage space, and the manufacturing footprint required for hydration.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidadConcentración
A44670BF200 L3 M
A44670BA5 L3 M
A44670BC20 L3 M
A44670BD50 L3 M
A44670BE100 L3 M
A65775BA5 L2 M
A65775BB20 L2 M
A65775BC50 L2 M
A65775BD100 L2 M
A65775BE200 L2 M
Número de catálogo A44670BF
Precio (CLP)
-
Cantidad:
200 L
Concentración:
3 M
Gibco concentrates are intended for dilution at the point of use and come sterile-filtered and packaged in conveniently sized bioprocess containers. Sourcing concentrated stock solutions can help streamline buffer preparation by helping reduce preparation time, storage space, and the manufacturing footprint required for hydration.
Features
  • Concentrated for dilution on-site
  • Available in a range of pack sizes
  • Established shelf life: set specifications
  • Made to order
  • Manufactured to ISO and cGMP standards to meet the stringent expectations of highly regulated industries
  • Readily accessible documentation to help accelerate qualification
Benefits
  • Helps ease time requirements for in-house buffer preparation
  • Mitigates space constraints for equipment storage
  • Streamlines in-house processes related to formulation, hydration, testing, and validation
  • Addresses workflow needs for biologics development and manufacturing stages
Additional information
  • Multi-compendial testing: compendial-grade or equivalent raw materials
  • Primary packaging: TAegis 5-14 single-use bioprocess container (BPC)
  • Secondary packaging: rigid shipping container with appropriate dunnage
  • Manufacturing facilities: ISO 13485–certified, cGMP-compliant to 21 CFR Part 820 standard
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
Concentración3 M
FormulaciónPhosphate Buffer Component (Acidic)
Calidad de fabricaciónCGMP
Cantidad200 L
FormularioClear, colorless solution
Tipo de productoSodium Phosphate Monobasic
EsterilidadSterile-filtered
Sterilization MethodSterile-filtered
Unit SizeEach