Commercial sale and application support for this item is provided by Floragenex, our global authorized distributor. Please contact Floragenex at agriseq.orders@floragenex.com for purchasing, pricing, and product inquiries.
Ion 530™ Chip Kit for Agrigenomics
Ion Torrent™
Ion 530™ Chip Kit for Agrigenomics
The Ion 530 Chip Kit contains 8 barcoded chips for sample tracking and sequencing with the Ion S5 and IonRead more
Have Questions?
Catalog NumberQuantity
A448738 chips
Catalog number A44873
Price (USD)
-
Quantity:
8 chips
The Ion 530 Chip Kit contains 8 barcoded chips for sample tracking and sequencing with the Ion S5 and Ion S5 XL Sequencing Systems. The Ion 530 Chip electronically detects polymerase-driven base incorporation without the use of fluorescence. By eliminating the use of an optical detection system, this advance in next-generation sequencing technology allows for rapid sequencing times of less than 2.5 hours for 200 bp sequencing.
The Ion 530 Chip offers:
• Sample tracking functionality from an integrated barcode
• A rapid run time of only 2.5–4 hours, generating 15–20 million reads
• Read lengths up to 600 bp to support a variety of research applications, including targeted gene sequencing and microbial sequencing
• Easy handling and loading of templated products for sequencing
• Compatibility with current library preparation methods
Notes:
This kit is only compatible with the Ion S5 and Ion S5 XL systems.
Use this kit with the Ion 510 & Ion 520 & Ion 530 Kit-Chef, the Ion 520 & Ion 530 ExT Kit-Chef, and the Ion 520 & Ion 530 Kit-OT2.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Ion GeneStudio S5 System, Ion S5 XL System
Number of Reads per Chip15-20 Million
Product TypeChip Kit
Quantity8 chips
Shipping ConditionRoom Temperature
Product LineIon 530
Unit SizeEach
Frequently asked questions (FAQs)
How many samples can I multiplex on a single chip for sequencing on the Ion PGM or Proton System?
The number of samples that can be multiplexed in a single sequencing run depends on the capacity of the chip, the size of the library, and the required coverage. A table of approximate capacities based on the size of the library and chip can be found in the Ion Ampliseq Preparation User Guide (https://tools.thermofisher.com/content/sfs/manuals/MAN0006735_AmpliSeq_DNA_RNA_LibPrep_UG.pdf) within the section “Strategies for combining Ion Ampliseq libraries.”
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What is the maximum throughput/day for the Ion S5 GeneStudio Prime System?
The Ion S5 GeneStudio Prime System has a maximum throughput/day of 50 Gb (2 Ion 550 Chips).
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How long does it take to run an Ion 550 Chip on the Ion S5 GeneStudio Prime System?
The Ion S5 GeneStudio Prime System takes 8.5 hours to run an Ion 550 Chip.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How long does it take to run an Ion 540 Chip on the Ion S5 GeneStudio Prime System?
The Ion S5 GeneStudio Prime System takes 6.5 hours to run an Ion 540 Chip.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What chips are compatible with the Ion GeneStudio S5 Prime System?
Ion 510 Chip, Ion 520 Chip, Ion 530 Chip, Ion 540 Chip, and Ion 550 Chip are compatible with the Ion GeneStudio S5 Prime System.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.