The Ion Torrent NGS Reverse Transcription (RT) Kit is the first cDNA synthesis kit developed specifically for next-generation sequencing (NGS)Más información
Have Questions?
Número de catálogo
Cantidad
A45003
32 reactions
Número de catálogo A45003
Precio (MXN)
3,929.01
Each
Cantidad:
32 reactions
Precio (MXN)
3,929.01
Each
The Ion Torrent NGS Reverse Transcription (RT) Kit is the first cDNA synthesis kit developed specifically for next-generation sequencing (NGS) applications. It combines the superior performance of SuperScript IV Reverse Transcriptase with a novel master mix optimized for NGS library preparation. The NGS RT Kit is compatible with Ion AmpliSeq and Ion AmpliSeq HD assays and can be used with both manual and automated workflows.
Benefits of the NGS RT Kit include: • Optimized for a variety of sample types, including FFPE, degraded samples, and those with low template amounts • Superior NGS results with novel RT formulation optimized for NGS library preparation • Helps increase NGS library yield, improve detection of long amplicons, and rescue under-performing amplicons • Helps maximize mapped reads and improve detection of RNA fusions • Trusted SuperScript IV RT technology offers superior cDNA synthesis performance with even the most challenging RNA samples • Universal kit for wide variety of Oncomine oncology research assays and Ion AmpliSeq panels
Simple reaction setup The NGS RT Kit contains a tube of 10X SuperScript IV enzyme mix and a tube of 5X buffer. A one-step, 25-minute reaction reverse transcribes total RNA into cDNA and can be used directly for NGS library preparation, without any additional quantification, purification, or dilution steps. A variety of sample types can be used, including RNA from blood, bone marrow, FFPE and partially degraded samples. This product is compatible with Oncomine and Ion AmpliSeq workflows, as well as Ion AmpliSeq HD panels. Consult the cDNA synthesis section of each assay manual for specific RT reaction volumes and setup instructions.
Performance and quality testing The NGS RT Kit is assayed for endodeoxyribonuclease, exodeoxyribonuclease, and ribonuclease, as well as yield and length of cDNA product. It is QC tested with a full NGS workflow to evaluate library yield and ensure performance meets key sequencing metrics.
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Concentración10X
Tipo de producto finalcDNA
Para utilizar con (aplicación)Next-Generation Sequencing, Reverse Transcription
Línea de productosIon Torrent
Tipo de productoReverse Transcription Kit
Cantidad32 reactions
Tipo de muestraRNA
Condiciones de envíoDry Ice
ObjetivoRNA
FormatoTube
Unit SizeEach
Contenido y almacenamiento
• 64 μL 5X Reaction Buffer, store at -5° to -30°C • 32 μL 10X RT Enzyme Mix, store at -5° to -30°C
Preguntas frecuentes
What are the shipping and storage conditions for the NGS Reverse Transcription Kit?
The NGS Reverse Transcription Kit is shipped on dry ice and we recommend storing all components at -30 degrees C to -10 degrees C (non-frost-free).
How many reactions can I perform with the NGS Reverse Transcription Kit?
The NGS Reverse Transcription Kit provides enough material to perform 32 reactions based on 10 µL reactions.
What is included in the NGS Reverse Transcription Kit?
The NGS Reverse Transcription Kit is the first cDNA synthesis kit developed specifically for next-generation sequencing (NGS) applications with specific validated assays. The kit contains 2 components:
- The 10X RT Enzyme Mix includes SuperScript IV Reverse Transcriptase, a proprietary RNase inhibitor, with helper proteins, and stabilizer proteins (1 tube x 32 µL)
- The 5X Reaction Buffer contains dNTPs, random hexamers, and MgCl2 in a formulation optimized for NGS libraries (1 tube x 64 µL)
How much of starting sample is needed for the NGS Reverse Transcription Kit?
We recommend using up to 2.5 µg of total RNA as starting material in a 10 µL reaction.
Note:
- Input amounts and reaction volumes vary with each assay and sample type. Consult assay-specific user guides for recommendations.
- To avoid PCR inhibition, RT reaction volume should be equal to or less than 50% of the PCR reaction.
Which sample types can I use with the NGS Reverse Transcription Kit?
A variety of sample types can be used with the NGS Reverse Transcription Kit, including RNA from blood, bone marrow, FFPE, and partially degraded samples. The NGS Reverse Transcription Kit is compatible with Ion AmpliSeq and Ion AmpliSeq HD assays and can be used with both manual and automated workflows.