Complete FTIR Library Collection
Thermo Scientific™
Complete FTIR Library Collection
The Complete FTIR Library Collection includes unlimited access for one user, for one full year, to more than 90,000 high-quality infrared spectra in high-resolution format.
Have Questions?
Catalog NumberProduct Line
A45128Thermo Scientific
Catalog number A45128
Price (USD)
0.00
Product Line:
Thermo Scientific
Price (USD)
0.00
The Complete FTIR Library Collection includes unlimited access for one user, for one full year, to more than 90,000 high-quality infrared spectra in high-resolution format. The collection includes hydrocarbons, alcohols, phenols, aldehydes, ketones, esters, anhydrides, lactones, dyes, indicators, alkynes, nitro and azo compounds, phosphorous- and sulphur-containing compounds, inorganics and silanes.
All libraries can be accessed using the search functionality included in the OMNIC Anywhere application available through our Connect cloud-based platform. Using data in your Connect account, your OMNIC data files can be quickly searched and results can be downloaded in several different file and report formats.
Subscriptions are purchased through ThermoFisher.com and delivered/managed through your Connect account. You will be notified of any changes to your subscription, including upcoming expiration dates.
Library updates are included with your subscription, so you will always have access to the latest spectral data.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineThermo Scientific
Quantity1 subscription
Unit SizeEach