QuantStudio™ 5 Spectral Calibration Plate (Cy™5.5 dye), 96-well, 0.1mL - FAQs
View additional product information for QuantStudio™ 5 Spectral Calibration Plate (Cy™5.5 dye), 96-well, 0.1mL - FAQs (A45218)
1 product FAQs found
Can I use the QuantStudio 5 Spectral Calibration Plate (Cy 5.5 dye), 96-well, 0.1mL (Cat. No. A45218) on the QuantStudio 5 System, 96-well, 0.2mL, instrument?
No, this would have to be ordered via our Specialty Oligos team (specialty_oligos@thermofisher.com).
Find additional tips, troubleshooting help, and resources within our QuantStudio 3 and QuantStudio 5 Real Time PCR Systems Support Center.