Why can I only buy the TrueCut Cas9 Protein (Prototype) today and not the full CTS product?
Real-time stability data is still being collected for full certification of the CTS product. During this period, we are making the same material available in prototype form, without full validation and real time stability information, to aid in preclinical development.