Oncomine™ BCR IGH LR Assay, RNA
Ion Torrent™

Oncomine™ BCR IGH LR Assay, RNA

The Ion Torrent Oncomine BCR IGH LR Assay is a robust, targeted next-generation sequencing (NGS) assay designed for use inRead more
Have Questions?
Catalog NumberNo. of Reactions
A4548524
Catalog number A45485
Price (USD)
7,790.00
Each
No. of Reactions:
24
Price (USD)
7,790.00
Each

The Ion Torrent Oncomine BCR IGH LR Assay is a robust, targeted next-generation sequencing (NGS) assay designed for use in hematology-oncology, immuno-oncology, infectious disease, and basic immunology research. Unlike other technologies, the Oncomine BCR IGH-LR Assay offers more than 400 base amplicons of library sequencing through long-read sequencing chemistry, enabling comprehensive coverage of the B-cell receptor (BCR) immunoglobin heavy (IGH) chain. The assay kit includes a single pool of multiplex PCR primers, cDNA synthesis kit, and Ion Ampliseq library reagents.

The Oncomine BCR IGH LR Assay is designed to accurately measure clonal expansion, quantify somatic hypermutation, and isotype B lymphocytes using total RNA extracted from bone marrow, whole blood, peripheral blood leukocytes (PBLs), peripheral blood mononuclear cells (PBMCs), or sorted cells in fresh-frozen research specimens. The assay utilizes Ion AmpliSeq multiplex PCR technology. It amplifies framework 1 (FR1) and the constant gene region of the BCR IGH gene to interrogate highly diverse complementarity-determining regions CDR1, CDR2, and CDR3, as well as the CH1 domain of the constant gene to enable distinction of all nine isotypes. IGH CDR-region amino acid motifs may reveal signatures of B-cell responses to defined antigens for use as future markers of autoimmune or infectious disease. Automated clonal lineage analysis and multi-sample analysis capability facilitates tracking of B-cell clonal evolution arising from somatic hypermutation and class switch recombination in research samples.

Oncomine BCR IGH LR Assay benefits include:

  • Extended amplicon allows for quantification of variable gene somatic hypermutation and isotype identification to enable hemato-oncology, immuno-oncology, and infectious disease translational and clinical research
  • Assay primers are designed to amplify all variable and joining gene alleles in the gold-standard IMGT database
  • RNA input improves detection of changes in plasmablast and plasma cell populations in research samples following immune challenge or administration of immunomodulatory agents
  • Superior multiplex PCR design through Ion AmpliSeq technology assures high accuracy and high sensitivity
  • Ultra-low substitution error rate minimizes artifacts arising from sequencing errors to enable highly accurate assessment of somatic hypermutation and clonal heterogeneity in malignant B-cell research samples of interest
  • The entire workflow from isolation of RNA to analysis of samples can be accomplished within two days using the Ion Chef templating system and Ion Genestudio S5 sequencing system
  • Flexible input requirements ensure successful library construction with as low as 25 ng and up to 2 μg total RNA input
  • Streamlined and user-friendly informatics solution offers automated clonotyping, somatic hypermutation quantification, clonal lineage analysis, reporting of key repertoire features, and multi-sample analysis capability to track B-cell clones across research samples

Learn more about the assay

Note: Information about data analysis using Ion Reporter Software v5.12 can be found in the User Guide below.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chip CompatibilityIon 520, Ion 530
For Use With (Application)Immune Repertoire Research
For Use With (Equipment)Ion GeneStudio S5, Ion GeneStudio S5 Plus, Ion GeneStudio S5 Prime, Ion Chef
LibrariesAmplicon
No. of Reactions24
No. of SamplesUp to 24 Samples
Nucleic Acid TypeRNA
Product LineOncomine
Product TypeAssay
Sample TypeWhole Blood, Fresh/Frozen Tissue, Sorted Cells
Shipping ConditionApproved for shipment on Wet or Dry Ice
ThroughputChip-dependent Multiplexing
Workflow StepPanel and Library
Sequencing TypeNext Generation Sequencing
Unit SizeEach
Contents & Storage
Store at -30°C to -10°C.

Frequently asked questions (FAQs)

How many libraries can be prepped with each Oncomine BCR IGH LR Assay, RNA?

The assay provides sufficient reagents for rapid preparation of 24 sample libraries.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

What is included in the Oncomine BCR IGH LR Assay, RNA?

The Oncomine BCR IGH LR Assay, RNA consists of the Oncomine BCR IGH-LR Panel, the Ion AmpliSeq Library Kit Plus (Cat. No. 4488990), and the Ion Torrent NGS Reverse Transcription Kit (Cat. No. A45003). Suffic’ent reagents are provided for the rapid preparation of 24 sample libraries.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

What is the input requirement for the Oncomine BCR IGH LR Assay, RNA?

The assay input requirement ranges from 25 ng to 2 µg DNase-treated total RNA. For RNA samples with a concentration of <6.25 ng/µL, we recommend using the DNAclear Purification Kit (Cat. No. AM1756) to concentrate the sample after performing reverse transcription. We recommend using ≥ 50 ng DNase-treated total RNA depending on sample type, ranging from 50-2,000 ng (see "Saturation analysis of peripheral blood leukocytes (PBL) - Oncomine BCR IGH Assay" on page 159 and "Saturation analysis of tissue - Oncomine BCR IGH Assay" on page 161 of the user guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0017438_OncomineTCRBetaAssay_UG.pdf).

When using degraded samples (FFPE) or samples with unknown B or T-cell content, perform sample qualification and quantitation to determine the minimum sample input that is required. For more information, see Appendix B, "Sample qualification and quantitation" on page 130 of the user guide.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

What types of samples is the Oncomine BCR IGH LR Assay, RNA compatible with?

This assay is compatible with isolated B cells, whole blood, peripheral blood leukocytes (PBLs), peripheral blood mononuclear cells (PBMCs), and fresh frozen tissue samples, including bone marrow samples.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

What is the Oncomine BCR IGH LR Assay, RNA?

The Oncomine BCR IGH LR Assay, RNA (Cat. No. A45485) is a highly sensitive, RNA-based NGS assay that enables the characterization of immunoglobulin heavy chain sequences, including all complementarity-determining regions (CDR1, 2, and 3) and the CH1 domain of the constant gene. The assay accurately measures B cell repertoire diversity and clonal expansion in a wide array of sample types. It allows for characterization of B cell isotype (and subtype), and the level of somatic hypermutation across the B cell receptor heavy chain.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.