Product Image
Applied Biosystems™
MagMAX™ DNA Ultra 2.0 with Cell and Tissue Extraction Buffer
This version of the MagMAX DNA Multi-Sample Ultra 2.0 Kit contains an additional buffer that allows you to extract DNA from tissues and cells as well as blood, saliva, buffy coat, and buccal swabs.
Have Questions?
Catalog NumberQuantity
A457211 Ea.
Catalog number A45721
Price (USD)
798.00
Each
Quantity:
1 Ea.
Price (USD)
798.00
Each
The MagMAX DNA Multi-Sample Ultra 2.0 Kit is an automatable magnetic bead-based sample preparation technology for the isolation of genomic DNA (gDNA) from 50 μL to 2 mL of whole blood, saliva, buffy coat, and buccal swabs. The resulting purified gDNA is ideal for a variety of downstream molecular biology applications such as qPCR, next-generation sequencing, and microarray analysis. One kit is sufficient to process 200 50–200 μL samples or 100 200–400 μL samples.
This version of the MagMAX DNA Multi-Sample Ultra 2.0 Kit contains an additional buffer that allows you to extract DNA from tissues and cells as well as blood, saliva, buffy coat, and buccal swabs.
Key features of the MagMAX DNA Multi-Sample Ultra 2.0 Kit:
  • Process 6–96 samples in 45 minutes with 5 minutes hands-on time
  • Isolate gDNA from 50 μL–2 mL of sample using a simple protocol in SBS microtiter plates
  • Skip sample and buffer volume normalization steps for hassle-free processing
  • Download validated BindIt software automation scripts for KingFisher Duo Prime and Flex purification systems
  • Automation-friendly stand-alone MagMAX DNA Multi-Sample Ultra 2.0 reagent bottle options available
Isolate genomic DNA from multiple sample types with a single sample prep kit
The MagMAX DNA Multi-Sample Ultra 2.0 Kit isolates gDNA from 50 μL–2 mL of whole blood (figure below), fresh and stabilized saliva (figure below), and EDTA/Streck cfDNA tube buffy coat (figure below) samples. Additionally, the same kit can be used to isolate gDNA from buccal swabs and other biological sample types. The MagMAX DNA Multi-Sample Ultra 2.0 Kit maximizes gDNA yields by treating samples with proteinase K (PK) and PK-enhancer followed by an innovative lysis/binding solution to simultaneously lyse cells, denature proteins, and capture gDNA on the high-binding capacity Ultra 2.0 gDNA magnetic binding beads. The Ultra 2.0 Wash I solution removes proteins, detergents, and other impurities from the bound nucleic acid. The gDNA is then released from the beads using a low-TE elution solution.
Fast and hassle-free MagMAX magnetic bead-based gDNA workflows
The MagMAX DNA Multi-Sample Ultra 2.0 Kit combines multiple laborious sample processing steps together into a single step to minimize the time and effort traditionally required by other methods without compromising yield and purity. Process a broad range of sample volume inputs simultaneously without sample normalization or reagent volume adjustments for hassle-free sample prep. Simply match your sample volumes to one of three standard input workflows (50 μL to 400 μL, up to 1 mL, up to 2 mL) and follow the work instructions for either manual or semi-automated sample preparation.
Made for KingFisher purification systems
The MagMAX DNA Multi-Sample Ultra 2.0 Kit is optimized for use on the moderate-throughput KingFisher Duo Prime (Cat. No. 5400110) and the higher-throughput KingFisher Flex (Cat. No. 5400630) purification systems. Users can process up to 400 μL of sample volume in high-throughput SBS 96-well deep-well microtiter plates using the Flex system. For smaller sample batches, users can process 12 samples using the Duo Prime system. Larger sample input volumes (1–2 mL), which are more challenging to process, are purified in SBS 24-well deep-well microtiter plates for either 24 samples on the Flex or 6 samples on the Duo Prime system. KingFisher instrument BindIt software scripts for all sample types and both Flex and Duo Prime systems can be downloaded below and installed onto the KingFisher instruments.
Working with higher throughput sample numbers?
The same MagMAX DNA Multi-Sample Ultra 2.0 Kit reagents are available as automation-friendly stand-alone reagents. MagMAX DNA Multi-Sample Ultra 2.0 stand-alone reagents are individually bottled in larger reagent volumes to make your high-thoughput workflows easier to handle. When purchased together as a sets of 6 stand-alone reagents, users will have sufficient volume of MagMAX DNA Multi-Sample Ultra 2.0 reagents to process 2000 50-200 μL samples or 1000 200-400 μL samples.
  • MagMAX DNA Ultra Multi-Sample 2.0 Proteinase K (45 mL) (Cat. No. A36578)
  • MagMAX DNA Ultra Multi-Sample 2.0 Binding Beads (45 mL) (Cat. No. A36579)
  • MagMAX DNA Ultra Multi-Sample 2.0 Wash I Solution (1.1 L) (Cat. No. A36580)
  • MagMAX DNA Ultra Multi-Sample 2.0 Lysis/Binding Solution (450 mL) (Cat. No. A36581)
  • MagMAX DNA Ultra Multi-Sample 2.0 Elution Solution (120 mL) (Cat. No. A36582)
  • MagMAX DNA Ultra Multi-Sample 2.0 Enhancer Solution (45 mL) (Cat. No. A36583)
For research use only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Next-Generation Sequencing, PCR, Real-Time PCR
High-throughput CompatibilityAutomated Protocols, High-throughput Compatible, Manual Protocols
Product LineMagMAX
Product TypeDNA Multi-Sample Ultra Kit
Quantity1 Ea.
Sample TypeCells, Tissue
Shipping ConditionRoom Temperature
TargetGenomic DNA
Unit SizeEach
Contents & Storage
• 1 MagMAX DNA Multi-Sample 2.0 Kit
• 60 mL Cell and Tissue DNA Extraction Buffer
Frequently asked questions (FAQs)
Can the MagMAX DNA Multi-Sample Ultra 2.0 Kit (Cat. No. A36570) be used to isolate genomic DNA from bone marrow aspirates and PBMCs?
Yes, we have protocols for using the MagMAX DNA Multi-Sample Ultra 2.0 Kit (Cat. No. A36570) to isolate genomic DNA from these sample types, but they require purchase of an additional Cell and Tissue Extraction Buffer. The kit and buffer can be purchased together as Cat. No. A45721 or the Cell and Tissue Extraction Buffer can be purchased as a stand alone item (Cat. No. A45469 or A45470).
The suggested workflow for bone marrow aspirates is as follows:
- Use only up to 200 µL of bone marrow aspirate
- Centrifuge at 200 x g for 10 mins
- Remove the supernatant (leaving about 50 µL of aspirate with the loose pellet) and resuspend the cell pellet in 400 µL of Cell and Tissue Extraction Buffer
- From this point, follow the protocol for isolation of DNA from buffy coat samples
The suggested workflow for 1-2.0 x 10E6 PBMCs is as follows:
- Centrifuge at 500-800 x g for 10 mins
- Remove the supernatant (leaving the cell pellet) an resuspend the cell pellet in 400 L of Cell and Tissue Extraction Buffer
- From this point, follow the protocol for isolation of DNA from cultured cell samples
Can the MagMAX DNA Multi-Sample Ultra 2.0 Kit (Cat. No. A36570) be used to isolate DNA from cultured cells and tissue?
Yes, we have protocols for using the MagMAX DNA Multi-Sample Ultra 2.0 Kit (Cat. No. A36570) to isolate DNA from cells and tissue, but they require purchase of an additional Cell and Tissue Extraction Buffer. The kit and buffer can be purchased together as Cat. No. A45721 or the Cell and Tissue Extraction Buffer can be purchased as a stand alone item (Cat. No. A45469 or A45470).
Do the MagMAX DNA Multi-Sample Ultra 2.0 cell and tissue DNA isolation protocols require an RNase A treatment step?
RNase A digestion is optional, but we highly recommend doing it. We recommend using PureLink RNase A (20 mg/mL) (Cat. No. 12091021).