This product ships via ground from US northeast.
Methanol, UHPLC-MS
Methanol, UHPLC-MS
Methanol, UHPLC-MS
Methanol, UHPLC-MS
Methanol, UHPLC-MS
Thermo Scientific™

Methanol, UHPLC-MS

Unique signal-to-noise specification for the chemical industry where the solvent quality is linked directly to the sensitivity of the mass spectrometer. | CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Catalog NumberQuantityPackaging
A45811 LClear Borosilicate Glass Bottle
Catalog number A4581
Price (USD)
70.65
Special offer
Online exclusive
Ends: 28-Dec-2026
82.39
Save 11.74
Each
Quantity:
1 L
Packaging:
Clear Borosilicate Glass Bottle
Price (USD)
70.65
Special offer
Online exclusive
Ends: 28-Dec-2026
82.39
Save 11.74
Each
Methanol, CH4O, CAS Number-67-56-1, wood naphtha, pyroxylic spirit, methyl alcohol, methylol, columbian spirit, colonial spirit, wood spirit, wood alcohol, methyl hydroxide, carbinol, 1L
RUO – Research Use Only
Specifications
Absorbance≤0.005 AU at 280nm, ≤0.01 AU at 254nm, ≤0.1 AU at 230nm, ≤0.4 AU at 215nm, ≤000 5 AU at 260nm, ≤02. AU at 220nm, ≤05. AU at 210nm
Assay Percent Range99.9% minimum by GC
Boiling Point64.7°C
CAS67-56-1
ChEBICHEBI:17790
Chemical Name or MaterialMethanol
ColorColorless
Density0.791 g/cm3
IUPAC Namemethanol
InChI KeyOKKJLVBELUTLKV-UHFFFAOYSA-N
Linear FormulaCH3OH
MDL NumberMFCD00004595
Melting Point-98°C
Molecular FormulaCH4O
PackagingClear Borosilicate Glass Bottle
Physical FormLiquid
PubChem CID887
Recommended StorageRoom temperature
Residue after Evaporation1ppm max.
SMILESCO
Synonymmethyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
Titratable Acid0.0003meq/g max.
Titratable Base0.0002meq/g max.
Vapor Pressure128 hPa at 20°C
Viscosity0.55 cP at 20°C
Water0.02% Max.
GradeUHPLC-MS
Molecular Weight (g/mol)32.04
Quantity1 L
Unit SizeEach