Ion 530™ Chip Kit-4 Reactions for Agrigenomics - FAQs
Ion 530™ Chip Kit-4 Reactions for Agrigenomics - FAQs
View additional product information for Ion 530™ Chip Kit-4 Reactions for Agrigenomics - FAQs (A45850)
1 product FAQs found
What is the throughput of the Ion 520 Chip?
For 200 bp sequencing, it will yield 0.6-1.0 Gb. For 400 bp sequencing, it will yield 1.2-2 Gb.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.