Product Image
Applied Biosystems™
SeqStudio™ Genetic Analyzer HID Upgrade
This package enables the upgrade of an existing RUO SeqStudio Genetic Analyzer to one that can be used for HID applications.
Have Questions?
Catalog NumberQuantity
A462301 instrument upgrade
Catalog number A46230
Price (USD)
-
Quantity:
1 instrument upgrade
This package enables the upgrade of an existing RUO SeqStudio Genetic Analyzer to one that can be used for HID applications. It provides access to SeqStudio DCS 1.2 and SAE V2.0 software and also includes an on-site visit by a field service engineer to upgrade the analyzer. Please note that HID applications also require a computer and GeneMapper ID-X 1.6 software, which are not included in this upgrade and may need to be purchased separately.
Specifications
For Use With (Equipment)Sequencing Instruments (Capillary Electrophoresis-Based)
Quantity1 instrument upgrade
Shipping ConditionRoom Temperature
Product LineSeqStudio
Unit SizeEach
Package Components
A46170
Catalog Number
A46170
Unit Size
Qty
1
A46169
Catalog Number
A46169
Unit Size
Qty
1
A46201
Catalog Number
A46201
Unit Size
Qty
1