Oncomine™ Comprehensive Assay v3 GX - FAQs
Oncomine™ Comprehensive Assay v3 GX - FAQs
View additional product information for Oncomine™ Comprehensive Assay v3 GX - FAQs (A46296)
5 product FAQs found
How many samples from the Oncomine Comprehensive Assay v3 GX can I run on a Genexus GX5 chip?
Each Genexus GX 5 chip can accommodate up to 6 DNA/RNA samples, or 5 samples and 1 no-template control.
What are the components of Oncomine Comprehensive Assay v3 GX (Cat. No. A46296)?
Oncomine Comprehensive Assay v3 GX (Cat. No. A46296) includes the following components:
- OCAv3 DNA Pool 1 (position 1, white cap) (4 tubes, 140 µL/tube), for 16 sequencing reactions (4 reactions per tube)
- OCAv3 DNA Pool 2 (position 1, white cap) (4 tubes, 140 µL/tube), for 16 sequencing reactions (4 reactions per tube)
- OCAv3 RNA Pool 1 (position 1, white cap) (4 tubes, 70 µL/tube), for 16 sequencing reactions (4 reactions per tube)
- OCAv3 RNA Pool 2 (position 1, white cap) (4 tubes, 70 µL/tube), for 16 sequencing reactions (4 reactions per tube)
- Genexus Strip 1 (2 x 8 strips), for 2 x 32 sequencing reactions (4 single‑pool reactions/strip)
- Genexus Strip 2-AS (2 x 8 strips), for 2 x 32 sequencing reactions (4 single‑pool reactions/strip)
Note: Genexus Library Strips 1 and 2-AS combo kit (Cat. No. A40252) can be purchased separately.
How much of each sample do I need for Oncomine Comprehensive Assay v3 GX?
Robust performance is observed from as little as 10 ng of nucleic acid input per pool (20 ng of DNA and 20 ng of RNA) isolated from FFPE samples, including fine needle biopsies.
What sample types can I use with Oncomine Comprehensive Assay v3 GX?
Oncomine Comprehensive Assay v3 GX is optimized for use with formalin-fixed paraffin embedded (FFPE) samples.
What is Oncomine Comprehensive Assay v3 GX (Cat. No. A46296) used for?
Oncomine Comprehensive Assay v3 GX (Cat. No. A46296) is a targeted, next-generation sequencing (NGS) assay that enables the detection of single-nucleotide variants (SNVs), insertions and deletions (INDELs), copy number variations (CNVs), and gene fusions from 161 unique genes. The assay is optimized for use with formalin-fixed paraffin embedded (FFPE) samples. The assay kit provides 4 pools of multiplex PCR primers (2 pools for the DNA assay and 2 pools for the RNA assay) and library reagents sufficient to perform up to sixteen 2-pool or eight 4-pool reactions on the Genexus Integrated Sequencer, using the Ion Genexus GX5 Chip.