Is the Plate Septa, 96 well (Cat. No. 4315933) compatible with 3500/3500xL Genetic Analyzers?
Yes, it is compatible with all versions of 3500/3500xL Genetic Analyzers as well as the 3500/3500xL Dx Genetic Analyzers, specifically in RUO mode.
Search
View additional product information for Applied Biosystems 3500 Dx Genetic Analyzer - FAQs (A46344)
3 product FAQs found