Applied Biosystems 3500xL Dx Genetic Analyzer - FAQs
Applied Biosystems 3500xL Dx Genetic Analyzer - FAQs
View additional product information for Applied Biosystems 3500xL Dx Genetic Analyzer - FAQs (A46345)
2 product FAQs found
Is the Plate Septa, 96 well (Cat. No. 4315933) compatible with 3500/3500xL Genetic Analyzers?
Yes, it is compatible with all versions of 3500/3500xL Genetic Analyzers as well as the 3500/3500xL Dx Genetic Analyzers, specifically in RUO mode.
Can a 3500 RUO Genetic Analyzer instrument be upgraded to a 3500 Dx IVD instrument?
A 3500 RUO RUO Genetic Analyzer instrument cannot be upgraded to a 3500 Dx IVD instrument because of differences in the regulated manufacturing processes for IVD instruments. However, a trade-in program is available and applies to all CE instruments.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.