You offer MicroSEQ ID 16S rDNA Full Gene Library v2.0, USB format (Cat. No. A46365) and MicroSEQ ID 16S rDNA Full Gene Library v2.0 (Cat. No. 4371295). What is the difference between them?
The only difference is that Cat. No. A46365 is available in USB format, otherwise the content is the same between the two catalog numbers.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.