Product Image
Applied Biosystems™
MicroSEQ™ ID Microbial Identification Software v3.1.3, Lite, with Software Library Bundle
This software bundle includes MicroSEQ ID Microbial Identification Software Lite v3.1.3, MicroSEQ ID 16S rDNA 500 Microbial Library, MicroSEQ IDRead more
Have Questions?
Catalog NumberQuantity
A465301 each
Catalog number A46530
Price (USD)
-
Quantity:
1 each
This software bundle includes MicroSEQ ID Microbial Identification Software Lite v3.1.3, MicroSEQ ID 16S rDNA 500 Microbial Library, MicroSEQ ID 16S rDNA Full Gene Library, and MicroSEQ ID Fungal Gene Library.
MicroSEQ ID Microbial Identification Software, Lite, version 3.1.3, allows you to easily identify and classify bacterial or fungal sequences by comparing them to a validated microbial library (available separately). Theis Lite version of the software is for use on a stand-alone PC (not connected to the genetic analyzer) for data analyses only.
Features of MicroSEQ ID Microbial Identification Software include:
• Automatically identify unknown specimens against a validated microbial library of over 3000 microbial sequences
• Run plates directly on Applied Biosystems 3500 Series genetic analyzers
• Equipped with security, audit, and e-signature features to enable 21 CFR Pt. 11 compliance
Confidently identify microbes
The MicroSEQ ID sequence libraries (available separately) consist of rigorously validated bacterial and fungal libraries to ensure accurate identification. The MicroSEQ ID 16S rDNA 500 Microbial Library consists of over 2300 500-bp 16S rDNA sequences, including extensive coverage of gram-negative non-fermenters, Bacillus, Coryneforms, Mycobacteria, and Staphylococcus. The entries contain the first 500 bases of the 16S rRNA gene, sufficient to identify bacterial unknowns.
The MicroSEQ ID Fungal Gene Library contains 1300 reference sequences from the D2 region of the Large Subunit (rDNA) of yeast and mold species against which an unknown yeast or mold sequence can be compared in order to identify the organism. The MicroSEQ libraries contain information on correctly identified organisms and quality-controlled sequences derived from type strains of curated culture collections. The libraries also support sequence polymorphisms and include bacterial Category A biothreat agents.
Automatically identify unknown specimens
Reviewing each specimen in a project file to make a manual identification call can be tedious and time consuming. The Auto-ID feature in MicroSEQ ID software uses a set of editable parameters to automatically identify unknown specimens. The Results table is displayed in the report with an editable Comments field for additional information or manual identification entries.
Run plates directly
Applied Biosystems 3500 Series genetic analyzer users are able to run plates directly via the MicroSEQ ID software user interface. A single software wizard enables you to set up projects, specimens, and plates; adjust the plate layout via drag and drop; start and monitor the run; and review results.
The MicroSEQ system: a complete, integrated solution
Streamlining every step in microbial identification, the MicroSEQ system combines the advantages of MicroSEQ ID software with easy-to-use PrepMan Ultra sample preparation reagents and protocols, MicroSEQ bacterial and fungal application kits, and industry-leading thermal cyclers and sequencing systems. To help ensure optimal performance and system-wide integration, we develop and test all components of the system together. MicroSEQ ID software also includes security, audit, and electronic signature features, enabling the software to smoothly integrate into your workflow.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)3500 Series Genetic Analyzers
Product LineMicroSEQ
Quantity1 each
Shipping ConditionRoom Temperature
System TypeCapillary Electrophoresis System Software
Product TypeMicrobial Identification Software
Unit SizeEach
Contents & Storage
• MicroSEQ ID Microbial Identification Software, version 3.1.3
• MicroSEQ ID 16S rDNA 500 Microbial Library
• MicroSEQ ID 16S rDNA Full Gene Library
• MicroSEQ ID Fungal Gene Library
Frequently asked questions (FAQs)
What is included in the MicroSEQ ID Microbial Identification Software v3.1.3, Lite, with Software Library Bundle (Cat. No. A46530)?
The components of the MicroSEQ ID Microbial Identification Software v3.1.3, Lite, with Software Library Bundle (Cat. No. A46530) are the following:
- MicroSEQ ID Microbial Identification Software, v3.1.3, Lite, WIN 10
- MicroSEQ ID Microbial Identification Software, v3.1.3, Lite License
- MicroSEQ ID Fungal Gene Library, v2018
- MicroSEQ ID Fungal Gene Library, v2018 License
- MicroSEQ ID 16S rDNA 500 Microbial Library, v2019
- MicroSEQ ID 16S rDNA 500 Microbial Library, v2019 License
- MicroSEQ ID 16S rDNA Full Gene Library
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What operating system is MicroSEQ ID Microbial Identification Software, Lite, version 3.1.3 supported on?
MicroSEQ ID Microbial Identification Software, Lite, version 3.1.3 is supported on Microsoft Windows 10, 64-bit operating system.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What are the recommended computer specifications for the MicroSEQ ID Microbial Identification Software, Lite, version 3.1.3?
Here are the recommended computer specifications for the MicroSEQ ID Microbial Identification Software, Lite, version 3.1.3:
- Processor: Intel Core i5 processor, 2.8 GHz or faster
- Hard drive: ≥250 GB for software installation and data storage
- Memory: ≥16 GB
- Operating System (OS): Windows 10 IOT, 64‑bit
IMPORTANT! We strongly recommend that you use computers that are provided by Thermo Fisher Scientific. All validation and verification studies were performed using Thermo Fisher Scientific computers.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What are the new features of the MicroSEQ ID Microbial Identification Software, Lite, version 3.1.3?
No new features were introduced into MicroSEQ ID Microbial Identification Software, Lite, version 3.1.3, compared to version 3.1. MicroSEQ ID Microbial Identification Software, Lite, version 3.1.3 is a stand-alone version of MicroSEQ ID Microbial Identification Software, version 3.1.3.
The Lite and System edition of MicroSEQ ID Microbial Identification Software are now compatible with 64-bit Windows 10 operating systems.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What is the MicroSEQ ID Microbial Identification Software, Lite, version 3.1.3 used for?
The MicroSEQ ID Microbial Identification Software, Lite, version 3.1.3 allows you to easily identify and classify bacterial or fungal sequences by comparing them to a validated microbial library (available separately). This Lite version of the software is for use on a stand-alone PC (not connected to the genetic analyzer) for data analyses only.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.