KingFisher Apex Purification Systems

KingFisher Apex Purification Systems

The KingFisher Apex Purification System is a versatile and user-friendly instrument that uses magnetic particle processing to automate the purification or isolation of DNA, RNA, proteins, and cells from virtually any source.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
A4694224 Combi Head, Ext Warranty
540091096 PCR Head
A4694196 PCR head, Ext Warranty
540092096 Combi head
A4694396 Combi head, Ext Warranty
540093096 DW head
A4694496 DW head, Ext Warranty
540094024 Combi head
Catalog number A46942
Price (USD)
-
Product Type:
24 Combi Head, Ext Warranty

The KingFisher Apex Purification System is a versatile and user-friendly instrument that uses magnetic particle processing to automate the purification or isolation of DNA, RNA, proteins, and cells from virtually any source. It moves magnetic beads, not liquids, through the main steps of sample preparation: bind, wash, and elute. This process provides excellent quality, reproducible results, and reduces hands-on time for sample processing.

  • User-friendly high throughput sample purification system for automating DNA/RNA, protein and cell purification
  • Provides versatile and high-speed purification with reproducible results
  • Easy to use interface with touch screen empowering the user to control all details of the application and purification process
  • BindIx PC Software for added convenience
  • Extensive selection of validated protocols for different type of applications
  • Cloud library for quick protocol retrieval
Specifications
Collection Efficiency of Particles>98% with 2.8 μm particle size
Cooling TemperatureDown to +4°C
Customer Education1 day at customer site
Depth (English)22 in.
Depth (Metric)56 cm
DisplayTouch screen (10 in.)
For Use With (Application)Genomics and proteomics, target identification, veterinary assays, biomarker discovery, quality control, forensics
Heating Temperature+4°C above ambient temperature up to +100°C
Height (English)18 in.
Height (Metric)46 cm
Magnetic Rods24 in one frame, interchangeable
Particle Size (Metric)ca. >1 μm
Plate Type24 DW, 24 storage tube
QuantitySystem + SmartStart + 1 yr ext warranty
Samples per Run24
Total Service Coverage1 year manufacturer warranty and 1 year of field service with PM
Type of Magnet Head24
UV Exposure TimeMax 23 hr. 59 min.
UV Lamp2 Pc.
Weight (English)123 lb.
Weight (Metric)56 kg
Width (English)31 in.
Width (Metric)79 cm
Capacity24 Samples
Format24-well
Plates per Deck8
Product Type24 Combi Head, Ext Warranty
Sample TypePlasma, Blood, Urine, Saliva, Cell Homogenate, Tissue Homogenate
Suitable Downstream AnalysisPCR/qPCR, Sequencing, Mass Spec
Volume (Metric) Processing24 DW plate 30 to 5000 μL, 24 storage tube 200 to 1000 μL
Unit SizeEach
Package Components
KingFisher Apex with 24 Combi head
Catalog Number
5400940
Unit Size
Each
Qty
1
ZG11SCKFAPEX
Catalog Number
ZG11SCKFAPEX
Unit Size
Qty
1
TRN46245
Catalog Number
TRN46245
Unit Size
Qty
1