RapidFinder™ Express Software v2.0 Kits
Thermo Scientific™

RapidFinder™ Express Software v2.0 Kits

Meet your pathogen and authenticity testing needs with this simple intuitive software enabling rapid, straightforward PCR detection.

Have Questions?
Catalog NumberFormat
A47006USB
Catalog number A47006
Price (TWD)
-
Format:
USB

Meet your pathogen and authenticity testing needs with the Thermo Scientific™ RapidFinder™ Express Software v2.0. This simple intuitive software enables rapid, straightforward PCR set-up, automatic data interpretation, and traceability of results.

RapidFinder™ Express Software v2.0 supports the Thermo Scientific™ SureTect™ PCR and Thermo Scientific™ RapidFinder™ Salmonella Multiplex PCR Assay workflows on the Applied Biosystems 7500 Fast Food Safety Real-Time PCR System.

The RapidFinder™ Express Software is a graphical, easy-to-use tool for pathogen detection applications. The software:

  • Leads you through the steps to specify the targets of interest and the number of samples and controls to run.
  • Gives you the flexibility to let the software determine the optimal location for unknown samples and controls on the plate or to customize the plate layout.
  • Leads you through the steps to start the instrument run and uses preset, optimized thermal cycling conditions for the selected assays.
  • Automatically generate the results (presence or absence of the pathogen). Results can be viewed for all locations, or for each sample, or for individual tubes.

The RapidFinder Express Software and the Applied Biosystems 7500 Fast Food Safety Real-Time PCR System are integral parts of the SureTect™ and RapidFinder™ Assays validated food pathogen testing workflows.

This product includes a Food and Environmental Real-Time PCR Analysis Software on the USB memory card, including release notes.

For Laboratory Use
For Laboratory Use

RapidFinder™ Express Software Version 2.0 is supplied on a credit card style USB drive, containing the files required for installation and release notes. Also includes SDS Software Version 1.4.2.1.

RapidFinder Express Software Version 2.0 supports use with Windows 7 and Windows 10 operating systems. If using Windows 10 then the version MUST be Windows 10 IOT 2019 to ensure correct operation of all functions.

Specifications
For Use With (Application)Sequencing
For Use With (Equipment)7500 Fast System
IncludesRapidFinder™ Express Software, Version 2.0, USB and release notes
Product LineRapidFinder
Product TypeExpress Software V2.0 Kit
Quantity1 Each
Shipping ConditionRoom Temperature
SoftwarePathogen Detection
FormatUSB
Unit SizeEach