MicroSEQ™ ID Microbial Identification Software, version 3.1.3 - FAQs
MicroSEQ™ ID Microbial Identification Software, version 3.1.3 - FAQs
View additional product information for MicroSEQ™ ID Microbial Identification Software, version 3.1.3 - FAQs (A47132)
3 product FAQs found
Does MicrobeBridge Software (Cat. No. A29238) require MicroSEQ ID Microbial Identification Software to be installed in order to function?
No, MicrobeBridge Software does not require MicroSeq ID Software to also be installed.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
Is the MicroSEQ ID Microbial Identification Software v3.0 Bundle for 3500 Instrument (3 licenses) (Cat. No. 4467661) compatible with Windows 10?
No, the only MicroSeq software that is compatible with Windows 10 is MicroSEQ ID Microbial Identification Software, version 3.1.3 (Cat. No. A47132).
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.