Learn more about financing options and programs » Learn more
QuantStudio™ 5 Dx Real-Time PCR System, desktop
Applied Biosystems™
QuantStudio™ 5 Dx Real-Time PCR System, desktop
The Applied Biosystems QuantStudio 5 Dx Real-Time PCR System is an in vitro diagnostic (IVD) quantitative PCR system that deliversRead more
Have Questions?
Catalog NumberQuantity
A473271 system
Catalog number A47327
Price (USD)
-
Quantity:
1 system
The Applied Biosystems QuantStudio 5 Dx Real-Time PCR System is an in vitro diagnostic (IVD) quantitative PCR system that delivers proven performance and support to help meet the needs of clinical/hospital laboratories and test developers, in a compact footprint and cost-effective package. This system is both US-IVD and CE-IVD, IVDR compliant.
Flexibility and security
The QuantStudio 5 Dx software is a combined-function software that enables both Development (DEV) and In Vitro Diagnostic (IVD) operation as discrete operating modes, minimizing risk of operation in the incorrect instance.
The software enables you to maintain secure access to the instrument, specify user access to various software functions, record all actions completed by users, and sign data electronically by virtue of Security, Auditing, and E-signature features.
This QuantStudio 5 Dx system is offered in a 96-well, 0.2 mL configuration and accommodates reaction volumes from 10 to 100 uL, as well as both standard and fast chemistries to meet varying user demands.
Trusted performance
Resulting from over 20 years of real-time PCR experience, the QuantStudio 5 Dx system includes proven Applied Biosystems OptiFlex technology and VeriFlex blocks for enhanced data quality. By enabling de-coupling of excitation and emission filters with OptiFlex technology, users may optimize performance for specific dyes and when multiplexing. With VeriFlex blocks, users may precisely set the temperature on six independently-controlled thermal zones across the 96-well sample block for optimal PCR amplification performance.
Cost effective
The QuantStudio 5 Dx system is our most affordable IVD qPCR platform. It is factory-calibrated and, following installation qualification and operational qualification by a factory-certified engineer, ready for operation by trained users. Subsequent IVD dye calibrations are now user-enabled for convenience and to minimize ongoing operating costs.
More information about the QuantStudio 5 Dx Real-Time PCR System ›
Learn more about available financing options and programs ›
For In Vitro Diagnostic Use.
Specifications
Dimensions40 x 27 x 50 cm (H x W x D)
Format96-well, 0.2 mL
Product LineQuantStudio
Quantity1 system
Shipping ConditionRoom Temperature
Weight27 kg
Block FormatNon-interchangeable
Unit SizeEach
Contents & Storage
• 1 QuantStudio 5 Dx 0.2 mL 96-well instrument
• 1 Dell tower computer with monitor and keyboard
• 1 QuantStudio 5 Dx IVD software kit
• 1 QuantStudio 5 Dx IVD user documentation kit
• 1 Installation and qualification service
Frequently asked questions (FAQs)
What is the difference between continuous melt curves and step and hold melt curves on the Applied Biosystems Real-Time PCR Instruments?
Continuous melt curve: increases the temperature by the ramp increment (degrees C/sec) so that the temperature will change at a constant rate set in the protocol.
Step and Hold melt curve: increases the temperature by the ramp increment (degrees C) and then holds at that temperature for the time specified by the user.
The amount of data collected will depend on the filter(s) selected. The more filters you use, the longer the acquisition will take and the fewer data points will be collected.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Can I start a run on the QuantStudio 5 Dx with the Development Software if the instrument calibrations are expired?
No. You cannot start a run on this instrument if calibrations are expired.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
What is the admin pin for QuantStudio 3 and 5 Real-Time PCR Systems?
If the admin PIN has not been changed, 1234 should work. If it does not work, you can also try 0000.
However, if the admin PIN has been changed and none of the above PINs work, follow the instructions in the QuantStudio 3 and 5 Real-Time PCR Systems manual, page 100.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
How do I download the Pathogen Interpretive Software?
Once you place your order, please contact our Instrument Service team at 1-800-955-6288 option 3, 1 (this kit is only available in North America). You will be given an access code to the TaqPath Monkeypox/Orthopox Virus DNA Kit eLearning where you’ll learn how to install, update, and download the Pathogen Interpretive Software package for the TaqPath Monkeypox/Orthopox Virus DNA Kit.
With the TaqPath Monkeypox/Orthopox Virus DNA Kit, what software should I use for data analysis?
Results are analyzed using the following software:
- Applied Biosystems Pathogen Interpretive Software v1.1
- SAE Administrator Console Dx v1.0 or v1.2 (for security and audit functions)
- Pathogen Interpretive Software 1.0.0 DAT (SAE Profile)
Please use the appropriate assay panel and template for your instrument:
- MPXOPX-EUA_QS5-9601_1.0.0.zip
- MPXOPX-EUA_QS5Dx-9602_1.0.0.zip
- MPXOPX-EUA_QS7F-384_1.0.0.zip
Note: These zip files are located in EducationConnect (www.thermofisher.com/educationconnect). The eLearning course needs to be completed to be granted access.