VeritiPro™ Thermal Cycler + Plastics Package, 96 well, EnduraPlates, 1 package - FAQs
VeritiPro™ Thermal Cycler + Plastics Package, 96 well, EnduraPlates, 1 package - FAQs
View additional product information for VeritiPro™ Thermal Cycler, 96-well - FAQs (A48766, A48767, A48768, A48769, A48141, A48770, A47397, A47396, A48771, A47398)
1 product FAQs found
Have you tested the Veriti and VeritiPro Thermal Cyclers in low humidity conditions (below 20%)?
The recommended conditions are determined to ensure no static discharge (low humidity) occurs.
Recommended Conditions:
- Veriti: 20-80% humidity
- VeritiPro: 15-80% humidity
We have not tested Veriti and VeritiPro thermal cyclers in low humidity, below 20%, therefore we cannot recommend using them in those conditions.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.