Hamilton™ CO-RE™ II Conductive Filter Tips, non-sterile, 300 μL
Thermo Scientific™
Hamilton™ CO-RE™ II Conductive Filter Tips, non-sterile, 300 μL
These filter tips are for use on the HID NIMBUS Presto System.Read more
Have Questions?
Catalog NumberQuantity
A476185760 Tips/Cs.
Catalog number A47618
Price (CLP)
-
Quantity:
5760 Tips/Cs.
These filter tips are for use on the HID NIMBUS Presto System.
Specifications
Certifications/ComplianceCertified for Biological Purity as DNase/RNase free and non-pyrogenic
DisposableSingle-use
Length (Metric)59.9 mm
Quantity5760 Tips/Cs.
AutoclavableNot Autoclavable
ColorBlack
FilteredFiltered
Fit for PurposeForensic Ready
For Use With (Equipment)HID Nimbus Presto
FormatBlister Refill
MaterialPolypropylene
Product LineHamilton CO-RE II
SterilityNon-sterile
Volume (Metric)300 μL
Unit SizeEach